Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06411948_cn
          See other PRKCA CNV Assays ›
          Gene Symbol
          PRKCA
          Assay Reference Genome
          Location

          Chr.17:66387375 on build GRCh38
          Cytoband
          17q24.2
          Assay ID Hs06411948_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5578

          Gene Name:

          protein kinase C alpha

          Gene Aliases:

          AAG6, PKC-alpha, PKCA, PRKACA

          Location:

          Chr.17:66302640-66810744 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PRKCA NM_002737.2 NP_002728.1
          XM_017024836.1 XP_016880325.1
          XM_017024841.1 XP_016880330.1
          AB209475.1 BAD92712.1
          AB451258.1
          AB451383.1
          AY633609.1 AAV33302.1
          BC053321.1 AAH53321.1
          BC109273.1 AAI09274.1
          BC109274.1 AAI09275.1
          LC151409.1
          M22199.1 AAA60098.1
          X52479.1 CAA36718.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv575894 Chr.17:66381708 - 66408004 on Build GRCh38 Loss PRKCA PRKCA-AS1
          nsv833520 Chr.17:66261176 - 66438257 on Build GRCh38 Gain PRKCA PRKCA-AS1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          positive regulation of endothelial cell proliferation
          desmosome assembly
          protein phosphorylation
          mitotic nuclear envelope disassembly
          cell adhesion
          activation of adenylate cyclase activity
          negative regulation of adenylate cyclase activity
          positive regulation of endothelial cell migration
          positive regulation of cardiac muscle hypertrophy
          peptidyl-serine phosphorylation
          platelet activation
          positive regulation of cell migration
          positive regulation of lipopolysaccharide-mediated signaling pathway
          negative regulation of glial cell apoptotic process
          histone H3-T6 phosphorylation
          intracellular signal transduction
          regulation of mRNA stability
          positive regulation of blood vessel endothelial cell migration
          positive regulation of macrophage differentiation
          positive regulation of angiogenesis
          positive regulation of cell adhesion
          positive regulation of mitotic cell cycle
          regulation of insulin secretion
          positive regulation of ERK1 and ERK2 cascade
          response to interleukin-1
          regulation of platelet aggregation
          apoptotic signaling pathway
          positive regulation of dense core granule biogenesis
          protein kinase activity
          protein serine/threonine kinase activity
          protein kinase C activity
          calcium-dependent protein kinase C activity
          protein binding
          ATP binding
          zinc ion binding
          enzyme binding
          histone kinase activity (H3-T6 specific)

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline