Search
Search

Entrez Gene ID: | 51755 |
Gene Name: | cyclin dependent kinase 12 |
Gene Aliases: |
CRK7, CRKR, CRKRS |
Location: |
Chr.17:39461486-39567559 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6498442 | Chr.17:39387510 - 39531174 on Build GRCh38 | Duplication |
|
| nsv1063848 | Chr.17:39419369 - 39551339 on Build GRCh38 | Gain |
|
Set Membership: |
|