Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06433409_cn
          See other OR3A2 CNV Assays ›
          Gene Symbol
          OR3A2
          Assay Reference Genome
          Location

          Chr.17:3280962 on build GRCh38
          Cytoband
          17p13.3
          Assay ID Hs06433409_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          4995

          Gene Name:

          olfactory receptor family 3 subfamily A member 2

          Gene Aliases:

          OLFRA04, OR17-14, OR17-228, OR228

          Location:

          Chr.17:3276113-3386311 on Build GRCh38

          Assay Gene Location:

          Within Intron 5
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          OR3A2 XM_017024697.1 XP_016880186.1
          XM_017024698.1 XP_016880187.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3892950 Chr.17:3131812 - 3577153 on Build GRCh38 Loss OR1A2 OR1D4 TRPV1 SPATA22 OR3A1 OR3A3 OR3A2 OR1E2 OR1A1 ASPA OR1E1 TRPV3 OR3A4P
          nsv1065489 Chr.17:2631416 - 3948349 on Build GRCh38 Gain OR1A2 CAMKK1 ITGAE CTNS OR1D4 TRPV1 SPATA22 MIR1253 LOC101927911 PAFAH1B1 OR3A3 OR3A2 OR1E2 OR1A1 ASPA NCBP3 OR1E1 TRPV3 P2RX1 OR1D2 MIR6776 CCDC92B RAP1GAP2 OR1D5 EMC6 GSG2 OR3A1 OR1G1 P2RX5-TAX1BP3 CLUH LOC105371592 LOC105371591 TAX1BP3 ATP2A3 OR3A4P SHPK P2RX5
          nsv574232 Chr.17:3254715 - 3503170 on Build GRCh38 Gain OR3A3 OR3A2 OR1E2 ASPA SPATA22 OR1E1 OR3A1 OR3A4P
          esv2660206 Chr.17:2679367 - 3519614 on Build GRCh38 Deletion OR1A2 OR1D2 OR1D4 MIR6776 CCDC92B SPATA22 MIR1253 LOC101927911 RAP1GAP2 OR1D5 PAFAH1B1 OR3A1 OR1G1 OR3A3 OR3A2 OR1E2 OR1A1 ASPA CLUH OR1E1 TRPV3 LOC105371592 LOC105371591 OR3A4P
          esv3639741 Chr.17:3279837 - 3291834 on Build GRCh38 Gain OR3A2 OR3A1
          esv3892949 Chr.17:3184567 - 3344389 on Build GRCh38 Loss OR1A2 OR3A2 OR1D4 OR1A1 OR3A1 OR3A4P
          nsv1060926 Chr.17:3093486 - 3319621 on Build GRCh38 Gain OR1A2 OR3A2 OR1D4 OR1A1 OR3A1 OR3A4P OR1G1
          nsv827859 Chr.17:3278220 - 3292037 on Build GRCh38 Gain OR3A2 OR3A1
          nsv574231 Chr.17:3208286 - 3290664 on Build GRCh38 Gain OR3A2 OR1D4 OR1A1 OR3A1
          nsv470570 Chr.17:3278090 - 3317250 on Build GRCh38 Gain OR3A2 OR3A1 OR3A4P

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of chemical stimulus
          detection of chemical stimulus involved in sensory perception of smell
          receptor activity
          G-protein coupled receptor activity
          olfactory receptor activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline