Search
Search

Entrez Gene ID: | 8490 |
Gene Name: | regulator of G-protein signaling 5 |
Gene Aliases: |
MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129 |
Location: |
Chr.1:163142299-163321791 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGS5 | NM_001254748.1 | NP_001241677.1 | ||
| NR_045630.1 | ||||
| BP292867.1 | ||||
| CB991106.1 |
Set Membership: |
|