Search
Search

Entrez Gene ID: | 1806 |
Gene Name: | dihydropyrimidine dehydrogenase |
Gene Aliases: |
DHP, DHPDHASE, DPD, DYPD |
Location: |
Chr.1:97077743-97921059 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPYD | NM_000110.4 | NP_000101.2 | ||
| XM_005270562.3 | XP_005270619.2 | |||
| XM_006710397.4 | XP_006710460.1 | |||
| XM_017000507.2 | XP_016855996.1 | |||
| XM_047448076.1 | XP_047304032.1 | |||
| XM_047448077.1 | XP_047304033.1 | |||
| AB003063.1 | BAA89789.1 | |||
| AK291217.1 | BAF83906.1 | |||
| BC131777.1 | AAI31778.1 | |||
| BC131778.1 | AAI31779.1 | |||
| U09178.1 | AAA57474.1 | |||
| U20938.1 | AAB51366.1 |
Entrez Gene ID: | 100873932 |
Gene Name: | DPYD antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:97095923-97322955 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPYD-AS1 | NR_046590.1 | |||
| CF529923.1 | ||||
| OA985456.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470728 | Chr.1:97237769 - 97296416 on Build GRCh38 | Loss |
|
Set Membership: |
|