Search
Search

Entrez Gene ID: | 4259 |
Gene Name: | microsomal glutathione S-transferase 3 |
Gene Aliases: |
GST-3, GST-III |
Location: |
Chr.1:165631234-165656136 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGST3 | NM_004528.4 | NP_004519.1 | ||
| XM_047421030.1 | XP_047276986.1 | |||
| AK312103.1 | BAG35039.1 | |||
| BC000505.2 | AAH00505.1 | |||
| BC005964.1 | AAH05964.1 | |||
| BI666647.1 | ||||
| DQ185045.1 | ABD14425.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4052465 | Chr.1:165642555 - 165653327 on Build GRCh38 | Deletion |
|
Set Membership: |
|