Search
Search

Entrez Gene ID: | 285362 |
Gene Name: | sulfatase modifying factor 1 |
Gene Aliases: |
AAPA3037, FGE, UNQ3037 |
Location: |
Chr.3:4034486-4467269 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3595104 | Chr.3:4418363 - 4483416 on Build GRCh38 | Loss |
|
| nsv6366184 | Chr.3:4456055 - 4460683 on Build GRCh38 | Deletion |
|
| nsv4085541 | Chr.3:4170096 - 4535684 on Build GRCh38 | Duplication |
|
| nsv5435055 | Chr.3:4418446 - 4483446 on Build GRCh38 | Deletion |
|
| nsv589451 | Chr.3:4231805 - 4473507 on Build GRCh38 | Loss |
|
| nsv6372572 | Chr.3:4402776 - 4526920 on Build GRCh38 | Deletion |
|
Set Membership: |
|