Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06656258_cn
          See other IRAK2 CNV Assays ›
          Gene Symbol
          IRAK2
          Assay Reference Genome
          Location

          Chr.3:10232508 on build GRCh38
          Cytoband
          3p25.3
          Assay ID Hs06656258_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          3656

          Gene Name:

          interleukin 1 receptor associated kinase 2

          Gene Aliases:

          IRAK-2

          Location:

          Chr.3:10164879-10243743 on Build GRCh38

          Assay Gene Location:

          Within Intron 10
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          IRAK2 NM_001570.3 NP_001561.3
          AF026273.1 AAB87669.1
          AJ496794.1 CAD43180.3
          AK299033.1
          AK312841.1
          BC125184.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv2662021 Chr.3:10231865 - 10239447 on Build GRCh38 Deletion IRAK2
          nsv436362 Chr.3:10048344 - 11860564 on Build GRCh38 Deletion LINC00606 LOC101927467 IRAK2 ATG7 TAMM41 HRH1 VGLL4 MIR885 MIR378B ATP2B2-IT2 SLC6A11 VHL FANCD2 SLC6A1 TATDN2 BRK1 GHRL FANCD2OS ATP2B2 SLC6A1-AS1 GHRLOS SEC13 LINC00852
          nsv821985 Chr.3:10231979 - 10239027 on Build GRCh38 Loss IRAK2
          dgv762e201 Chr.3:10008967 - 11913136 on Build GRCh38 Deletion LOC101927467 HRH1 VGLL4 MIR885 MIR378B ATP2B2-IT2 SLC6A11 VHL FANCD2 SLC6A1 BRK1 ATP2B2 GHRLOS SEC13 LINC00606 LOC401052 IRAK2 ATG7 TAMM41 EMC3 CIDECP TATDN2 GHRL FANCD2OS FANCD2P2 SLC6A1-AS1 LINC00852
          nsv1121603 Chr.3:10071433 - 11889279 on Build GRCh38 Deletion LINC00606 LOC101927467 IRAK2 ATG7 TAMM41 HRH1 VGLL4 MIR885 MIR378B ATP2B2-IT2 SLC6A11 VHL FANCD2 SLC6A1 TATDN2 BRK1 GHRL FANCD2OS FANCD2P2 ATP2B2 SLC6A1-AS1 GHRLOS SEC13 LINC00852
          nsv436869 Chr.3:10064456 - 11877985 on Build GRCh38 Insertion LINC00606 LOC101927467 IRAK2 ATG7 TAMM41 HRH1 VGLL4 MIR885 MIR378B ATP2B2-IT2 SLC6A11 VHL FANCD2 SLC6A1 TATDN2 BRK1 GHRL FANCD2OS FANCD2P2 ATP2B2 SLC6A1-AS1 GHRLOS SEC13 LINC00852
          nsv522516 Chr.3:10195263 - 10236182 on Build GRCh38 Gain IRAK2
          nsv1073204 Chr.3:9996548 - 11875005 on Build GRCh38 Deletion LOC101927467 EMC3-AS1 HRH1 VGLL4 MIR885 MIR378B ATP2B2-IT2 SLC6A11 VHL FANCD2 SLC6A1 BRK1 ATP2B2 GHRLOS SEC13 LINC00606 LOC401052 IRAK2 ATG7 TAMM41 EMC3 CIDECP TATDN2 GHRL FANCD2OS FANCD2P2 SLC6A1-AS1 LINC00852
          esv3595271 Chr.3:10231881 - 10239570 on Build GRCh38 Loss IRAK2

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          regulation of cytokine-mediated signaling pathway
          toll-like receptor signaling pathway
          MyD88-dependent toll-like receptor signaling pathway
          protein phosphorylation
          inflammatory response
          I-kappaB kinase/NF-kappaB signaling
          JNK cascade
          lipopolysaccharide-mediated signaling pathway
          negative regulation of NF-kappaB transcription factor activity
          toll-like receptor 9 signaling pathway
          positive regulation of NF-kappaB transcription factor activity
          nucleotide-binding oligomerization domain containing signaling pathway
          interleukin-1-mediated signaling pathway
          response to interleukin-1
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          protein homodimerization activity
          protein heterodimerization activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline