Search
Search

Entrez Gene ID: | 2632 |
Gene Name: | 1,4-alpha-glucan branching enzyme 1 |
Gene Aliases: |
APBD, GBE, GSD4 |
Location: |
Chr.3:81489703-81761645 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBE1 | NM_000158.4 | NP_000149.4 | ||
| AB209731.1 | BAD92968.1 | |||
| AK125918.1 | BAG54265.1 | |||
| AK297939.1 | BAG60255.1 | |||
| AK300620.1 | BAG62313.1 | |||
| AK312245.1 | ||||
| BC012098.1 | AAH12098.1 | |||
| L07956.1 | AAA58642.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4083598 | Chr.3:81537753 - 81686872 on Build GRCh38 | Deletion |
|
| nsv5993998 | Chr.3:81554863 - 81565508 on Build GRCh38 | Duplication |
|
Set Membership: |
|