Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06733459_cn
          See other E2F3 CNV Assays ›
          Gene Symbol
          E2F3
          Assay Reference Genome
          Location

          Chr.6:20406064 on build GRCh38
          Cytoband
          6p22.3
          Assay ID Hs06733459_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          1871

          Gene Name:

          E2F transcription factor 3

          Gene Aliases:

          E2F-3

          Location:

          Chr.6:20353285-20493714 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          E2F3 NM_001243076.2 NP_001230005.1
          NM_001949.4 NP_001940.1
          XM_005248865.4 XP_005248922.1
          XM_011514324.2 XP_011512626.1
          BC099908.1 AAH99908.1
          BC114554.1 AAI14555.1
          CR749285.1 CAH18140.1
          Y10479.1 CAA71504.1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic non-DGV Variation

          Panther Classification:

          Molecular Function -

          RNA metabolism protein nucleic acid metabolism protein general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription initiation from RNA polymerase II promoter
          cell cycle
          positive regulation of cell proliferation
          positive regulation of transcription, DNA-templated
          negative regulation of fat cell proliferation
          core promoter binding
          transcription factor activity, sequence-specific DNA binding
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline