Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06811326_cn
          See other GCNT2 CNV Assays ›
          Gene Symbol
          GCNT2
          Assay Reference Genome
          Location

          Chr.6:10620312 on build GRCh38
          Cytoband
          Assay ID Hs06811326_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          2651

          Gene Name:

          glucosaminyl (N-acetyl) transferase 2 (I blood group)

          Gene Aliases:

          CCAT, CTRCT13, GCNT2C, GCNT5, IGNT, II, NACGT1, NAGCT1, ULG3, bA360O19.2, bA421M1.1

          Location:

          Chr.6:32235-140252 on Build GRCh38

          Assay Gene Location:

          Within Intron 7
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          GCNT2 NM_001374747.1 NP_001361676.1
          NM_001491.3 NP_001482.1
          NM_145649.5 NP_663624.1
          NM_145655.4 NP_663630.2
          XM_005248999.3 XP_005249056.1
          XM_006715052.4 XP_006715115.1
          XM_047418633.1 XP_047274589.1
          AB078432.1 BAC66781.1
          AB078433.1 BAC66782.1
          AF401652.1 AAL50561.1
          AF401653.1 AAL50562.1
          AF458024.1 AAM73864.1
          AF458025.1 AAM73865.1
          AF458026.1 AAM73866.1
          AK090483.1 BAC03464.1
          AK098105.1 BAC05234.1
          AK291195.1 BAF83884.1
          AK291767.1 BAF84456.1
          AK307329.1
          AK307337.1
          AK307373.1
          AK313426.1 BAG36218.1
          AK313903.1 BAG36626.1
          AL832714.1
          AL832719.1
          AY435145.1 AAR95646.1
          AY435146.1 AAR95647.1
          AY435147.1 AAR95648.1
          BC074801.2 AAH74801.1
          BC074802.2 AAH74802.1
          BC130524.1 AAI30525.1
          BX647576.1 CAI46081.1
          BX647668.1
          L19659.1 AAA81777.1
          Z19550.1 CAA79610.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv3113907 Chr.6:10481673 - 10698466 on Build GRCh38 Deletion GCNT2 C6orf52 LINC02522 PAK1IP1
          nsv966670 Chr.6:10447438 - 10871656 on Build GRCh38 Duplication GCNT2 TMEM14C TMEM14B C6orf52 MIR5689HG LINC02522 PAK1IP1 MAK TMEM14B-DT
          nsv600979 Chr.6:10449025 - 10839526 on Build GRCh38 Gain GCNT2 TMEM14C TMEM14B C6orf52 MIR5689HG LINC02522 PAK1IP1 MAK TMEM14B-DT
          nsv6404362 Chr.6:10545723 - 10821818 on Build GRCh38 Duplication GCNT2 TMEM14C TMEM14B C6orf52 PAK1IP1 MAK TMEM14B-DT
          nsv470800 Chr.6:10538711 - 10863092 on Build GRCh38 Gain GCNT2 TMEM14C TMEM14B C6orf52 PAK1IP1 MAK TMEM14B-DT
          nsv6403222 Chr.6:10465448 - 10718419 on Build GRCh38 Deletion GCNT2 C6orf52 LINC02522 PAK1IP1
          esv2759406 Chr.6:10443564 - 10642899 on Build GRCh38 Loss GCNT2 MIR5689HG LINC02522

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          glycosaminoglycan biosynthetic process
          protein glycosylation
          transforming growth factor beta receptor signaling pathway
          multicellular organism development
          positive regulation of cell proliferation
          glycoprotein biosynthetic process
          posttranscriptional regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          negative regulation of cell-substrate adhesion
          positive regulation of cell migration
          positive regulation of heterotypic cell-cell adhesion
          maintenance of lens transparency
          positive regulation of protein kinase B signaling
          positive regulation of ERK1 and ERK2 cascade
          protein binding
          N-acetyllactosaminide beta-1,6-N-acetylglucosaminyltransferase activity
          UDP-glycosyltransferase activity
          acetylglucosaminyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline