Search
Search

Entrez Gene ID: | 4216 |
Gene Name: | mitogen-activated protein kinase kinase kinase 4 |
Gene Aliases: |
MAPKKK4, MEKK 4, MEKK4, MTK1, PRO0412 |
Location: |
Chr.6:160991521-161117385 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K4 | NM_001291958.1 | NP_001278887.1 | ||
| NM_001301072.1 | NP_001288001.1 | |||
| NM_005922.3 | NP_005913.2 | |||
| NM_006724.3 | NP_006715.2 | |||
| NR_120425.1 | ||||
| XM_005266989.2 | XP_005267046.1 | |||
| XM_017010869.1 | XP_016866358.1 | |||
| AF002715.1 | AAB68804.1 | |||
| AK307866.1 | ||||
| BC136276.1 | ||||
| BC143735.1 | ||||
| BC146770.1 | ||||
| D86968.2 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3611453 | Chr.6:160930783 - 161147021 on Build GRCh38 | Gain |
|
| esv3576242 | Chr.6:160924181 - 161141222 on Build GRCh38 | Gain |
|
| nsv1030045 | Chr.6:160938043 - 161141223 on Build GRCh38 | Gain |
|
| nsv1026543 | Chr.6:160306836 - 161585804 on Build GRCh38 | Gain |
|
Set Membership: |
|