Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06860795_cn
          See other MTAP CNV Assays ›
          Gene Symbol
          MTAP
          Assay Reference Genome
          Location

          Chr.9:21806692 on build GRCh38
          Cytoband
          Assay ID Hs06860795_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          4507

          Gene Name:

          methylthioadenosine phosphorylase

          Gene Aliases:

          BDMF, DMSFH, DMSMFH, HEL-249, LGMBF, MSAP, c86fus

          Location:

          Chr.9:21802636-21941115 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          MTAP NM_001396040.1 NP_001382969.1
          NM_001396041.1 NP_001382970.1
          NM_001396042.1 NP_001382971.1
          NM_001396043.1 NP_001382972.1
          NM_001396044.1 NP_001382973.1
          NM_001396045.1 NP_001382974.1
          NM_002451.4 NP_002442.2
          NR_173242.1
          AF109294.1 AAD19641.1
          AK300592.1 BAG62290.1
          AK309365.1
          BC012316.1 AAH12316.1
          BC026106.1 AAH26106.1
          CR541670.1 CAG46471.1
          CR541710.1 CAG46511.1
          FJ224338.1 ACI46030.1
          GQ891397.1 ADO22259.1
          HE654772.1 CCF77345.1
          HE654773.1 CCF77346.1
          HE654774.1 CCF77347.1
          HE654775.1 CCF77348.1
          HE654776.1 CCF77349.1
          HE654777.1 CCF77350.1
          L40432.1 AAG38871.1
          U22233.1 AAA81646.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv4184515 Chr.9:21703459 - 22598303 on Build GRCh38 Duplication DMRTA1 MTAP CDKN2A CDKN2B-AS1 CDKN2A-AS1 CDKN2B
          nsv6000594 Chr.9:21677724 - 23021905 on Build GRCh38 Duplication DMRTA1 MTAP CDKN2A LINC01239 CDKN2B-AS1 CDKN2A-AS1 CDKN2B
          nsv1026981 Chr.9:21748203 - 21811698 on Build GRCh38 Gain MTAP
          esv2764150 Chr.9:20659506 - 22349184 on Build GRCh38 Gain+Loss HACD4 IFNA22P KLHL9 FOCAD-AS1 IFNA14 MIR491 IFNA21 FOCAD IFNA5 IFNA17 IFNA16 MTAP CDKN2A IFNB1 IFNA13 IFNA10 CDKN2B-AS1 IFNE MIR31HG IFNA2 IFNA4 MIR31 IFNW1 IFNA8 CDKN2A-AS1 IFNA7 IFNA1 CDKN2B SNORA30B IFNA6
          nsv8426 Chr.9:21763811 - 22176201 on Build GRCh38 Loss MTAP CDKN2A CDKN2B-AS1 CDKN2A-AS1 CDKN2B
          esv3619972 Chr.9:21806006 - 21817430 on Build GRCh38 Loss MTAP
          nsv5479271 Chr.9:21806005 - 21817449 on Build GRCh38 Deletion MTAP

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleobase-containing compound metabolic process
          purine ribonucleoside salvage
          nicotinamide riboside catabolic process
          nucleoside metabolic process
          L-methionine biosynthetic process from methylthioadenosine
          methylation
          response to testosterone
          catalytic activity
          phosphorylase activity
          protein binding
          transferase activity
          transferase activity, transferring glycosyl groups
          transferase activity, transferring pentosyl groups
          S-methyl-5-thioadenosine phosphorylase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline