Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06867558_cn
          See other LPAR1 CNV Assays ›
          Gene Symbol
          LPAR1
          Assay Reference Genome
          Location

          Chr.9:111015831 on build GRCh38
          Cytoband
          9q31.3
          Assay ID Hs06867558_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          1902

          Gene Name:

          lysophosphatidic acid receptor 1

          Gene Aliases:

          EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1

          Location:

          Chr.9:110873252-111039259 on Build GRCh38

          Assay Gene Location:

          Within Intron 7
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          LPAR1 NM_001401.3 NP_001392.2
          NM_057159.2 NP_476500.1
          XM_005251781.3 XP_005251838.1
          XM_005251782.3 XP_005251839.1
          XM_017014383.1 XP_016869872.1
          XM_017014384.1 XP_016869873.1
          XM_017014385.1 XP_016869874.1
          XM_017014386.1 XP_016869875.1
          XM_017014387.1 XP_016869876.1
          XM_017014388.1 XP_016869877.1
          XM_017014389.1 XP_016869878.1
          XM_017014390.1 XP_016869879.1
          XM_017014391.1 XP_016869880.1
          XM_017014392.1 XP_016869881.1
          XM_017014393.1 XP_016869882.1
          XM_017014394.1 XP_016869883.1
          XM_017014395.1 XP_016869884.1
          XM_017014396.1 XP_016869885.1
          XM_017014397.1 XP_016869886.1
          XM_017014398.1 XP_016869887.1
          XM_017014399.1 XP_016869888.1
          XM_017014400.1 XP_016869889.1
          XM_017014401.1 XP_016869890.1
          XM_017014402.1 XP_016869891.1
          XM_017014403.1 XP_016869892.1
          XM_017014405.1 XP_016869894.1
          XM_017014406.1 XP_016869895.1
          XM_017014407.1 XP_016869896.1
          AK022808.1
          AK290287.1
          AK298590.1
          AK299279.1
          BC036034.1 AAH36034.1
          BC073167.1 AAH73167.1
          BI461641.1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic non-DGV Variation

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          G-protein coupled receptor signaling pathway
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          activation of phospholipase C activity
          positive regulation of cytosolic calcium ion concentration
          regulation of cell shape
          negative regulation of neuron projection development
          oligodendrocyte development
          cerebellum development
          optic nerve development
          corpus callosum development
          negative regulation of cAMP biosynthetic process
          bleb assembly
          positive regulation of Rho protein signal transduction
          myelination
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of MAPK cascade
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
          positive regulation of stress fiber assembly
          cell chemotaxis
          positive regulation of dendritic spine development
          cellular response to oxygen levels
          positive regulation of smooth muscle cell chemotaxis
          cellular response to 1-oleoyl-sn-glycerol 3-phosphate
          G-protein alpha-subunit binding
          G-protein coupled receptor activity
          protein binding
          PDZ domain binding
          lysophosphatidic acid binding
          lysophosphatidic acid receptor activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline