Search
Search

Entrez Gene ID: | 9380 |
Gene Name: | glyoxylate and hydroxypyruvate reductase |
Gene Aliases: |
GLXR, GLYD, PH2 |
Location: |
Chr.9:37422435-37439494 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRHPR | NM_012203.2 | NP_036335.1 | ||
| XM_005251631.2 | XP_005251688.1 | |||
| XM_017015320.3 | XP_016870809.1 | |||
| XM_017015321.3 | XP_016870810.1 | |||
| XM_024447716.2 | XP_024303484.1 | |||
| XM_047424084.1 | XP_047280040.1 | |||
| AF113215.1 | AAG39286.1 | |||
| AF113251.1 | AAD54066.1 | |||
| AF134895.1 | AAF00111.1 | |||
| AF146018.1 | AAD45886.1 | |||
| AK025978.1 | ||||
| AK307325.1 | ||||
| AK307902.1 | ||||
| AK315690.1 | BAG38053.1 | |||
| BC000605.2 | AAH00605.1 | |||
| BC088360.1 | AAH88360.1 | |||
| GQ891362.1 | ADO22224.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5480716 | Chr.9:37423745 - 37424780 on Build GRCh38 | Deletion |
|
Set Membership: |
|