Search
Search

Entrez Gene ID: | 53919 |
Gene Name: | solute carrier organic anion transporter family member 1C1 |
Gene Aliases: |
OATP-F, OATP1, OATP14, OATP1C1, OATPF, OATPRP5, SLC21A14 |
Location: |
Chr.12:20690505-20753386 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3628788 | Chr.12:20691619 - 20734169 on Build GRCh38 | Gain |
|
| nsv1054997 | Chr.12:20529075 - 21139668 on Build GRCh38 | Gain |
|
| nsv557704 | Chr.12:20689107 - 20741046 on Build GRCh38 | Gain |
|
| nsv1159988 | Chr.12:20604634 - 20741072 on Build GRCh38 | Duplication |
|
Set Membership: |
|