Search
Search

Entrez Gene ID: | 55500 |
Gene Name: | ethanolamine kinase 1 |
Gene Aliases: |
EKI, EKI 1, EKI1, Nbla10396 |
Location: |
Chr.12:22625142-22690674 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ETNK1 | NM_001039481.1 | NP_001034570.1 | ||
| NM_018638.4 | NP_061108.2 | |||
| XM_005253420.4 | XP_005253477.1 | |||
| XM_017019580.1 | XP_016875069.1 | |||
| XM_017019581.1 | XP_016875070.1 | |||
| AF207600.2 | AAF71220.2 | |||
| AK074425.1 | ||||
| AL137750.1 | CAH56371.1 | |||
| BC006111.1 | ||||
| BC066907.1 | AAH66907.1 | |||
| BT007084.1 | AAP35747.1 |
Set Membership: |
|