Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07127080_cn
          See other PTPRS CNV Assays ›
          Gene Symbol
          PTPRS
          Assay Reference Genome
          Location

          Chr.19:5224939 on build GRCh38
          Cytoband
          Assay ID Hs07127080_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5802

          Gene Name:

          protein tyrosine phosphatase receptor type S

          Gene Aliases:

          PTP-sigma, PTPSIGMA, R-PTP-S, R-PTP-sigma

          Location:

          Chr.19:5205508-5340812 on Build GRCh38

          Assay Gene Location:

          Within Intron 20
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PTPRS NM_001394011.1 NP_001380940.1
          NM_001394012.1 NP_001380941.1
          NM_001394013.1 NP_001380942.1
          NM_002850.4 NP_002841.3
          NM_130853.3 NP_570923.2
          NM_130854.3 NP_570924.2
          NM_130855.3 NP_570925.2
          XM_005259600.3 XP_005259657.1
          XM_005259607.3 XP_005259664.1
          XM_005259610.2 XP_005259667.1
          XM_011528158.3 XP_011526460.1
          XM_017027065.2 XP_016882554.1
          XM_017027066.2 XP_016882555.1
          XM_017027067.2 XP_016882556.1
          XM_017027068.2 XP_016882557.1
          XM_017027069.2 XP_016882558.1
          XM_017027070.2 XP_016882559.1
          XM_017027071.2 XP_016882560.1
          XM_017027072.2 XP_016882561.1
          XM_017027073.2 XP_016882562.1
          XM_017027074.2 XP_016882563.1
          XM_017027075.2 XP_016882564.1
          XM_047439155.1 XP_047295111.1
          XM_047439156.1 XP_047295112.1
          XM_047439157.1 XP_047295113.1
          XM_047439158.1 XP_047295114.1
          XM_047439159.1 XP_047295115.1
          XM_047439160.1 XP_047295116.1
          XM_047439161.1 XP_047295117.1
          XM_047439162.1 XP_047295118.1
          XM_047439163.1 XP_047295119.1
          XM_047439164.1 XP_047295120.1
          XM_047439165.1 XP_047295121.1
          XM_047439166.1 XP_047295122.1
          XM_047439167.1 XP_047295123.1
          AB209333.1 BAD92570.1
          BC104812.1 AAI04813.1
          BC143287.1 AAI43288.1
          U35234.1 AAC50299.1
          U40317.1 AAC50567.1
          U41725.1 AAD09360.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv833723 Chr.19:5106817 - 5273455 on Build GRCh38 Loss PTPRS-AS1 PTPRS KDM4B
          nsv953951 Chr.19:5065290 - 5240089 on Build GRCh38 Deletion PTPRS-AS1 PTPRS KDM4B
          nsv833724 Chr.19:5213508 - 5399018 on Build GRCh38 Loss LOC105372253 PTPRS
          nsv510750 Chr.19:5121816 - 5246204 on Build GRCh38 Deletion PTPRS-AS1 PTPRS KDM4B

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          cell adhesion
          signal transduction
          negative regulation of neuron projection development
          spinal cord development
          cerebellum development
          hippocampus development
          cerebral cortex development
          corpus callosum development
          negative regulation of axon extension
          negative regulation of interferon-alpha production
          negative regulation of interferon-beta production
          negative regulation of toll-like receptor 9 signaling pathway
          peptidyl-tyrosine dephosphorylation
          negative regulation of collateral sprouting
          negative regulation of axon regeneration
          modulation of synaptic transmission
          synapse organization
          negative regulation of dendritic spine development
          establishment of endothelial intestinal barrier
          regulation of postsynaptic density assembly
          trans-synaptic signaling
          synaptic membrane adhesion
          phosphoprotein phosphatase activity
          protein tyrosine phosphatase activity
          protein binding
          heparin binding
          hydrolase activity
          chondroitin sulfate binding
          heparan sulfate proteoglycan binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline