Search
Search

Entrez Gene ID: | 128826 |
Gene Name: | MIR1-1 host gene |
Gene Aliases: |
C20orf166, MIR133A2HG |
Location: |
Chr.20:62550453-62570764 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR1-1HG | NR_171007.1 | |||
| AK056985.1 | ||||
| BC110595.2 | AAI10596.1 | |||
| DA896197.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422398 | Chr.20:62207833 - 62674040 on Build GRCh38 | Duplication |
|
| dgv4344n100 | Chr.20:62436472 - 62569333 on Build GRCh38 | Gain |
|
| nsv586496 | Chr.20:62332390 - 62812195 on Build GRCh38 | Gain |
|
| nsv953302 | Chr.20:62345345 - 62831248 on Build GRCh38 | Deletion |
|
| nsv459068 | Chr.20:62444168 - 62577283 on Build GRCh38 | Gain |
|
| nsv470556 | Chr.20:62212004 - 62577283 on Build GRCh38 | Loss |
|
| dgv4343n100 | Chr.20:62391262 - 62569333 on Build GRCh38 | Gain |
|