Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07207836_cn
          See other BMP7 CNV Assays ›
          Gene Symbol
          BMP7
          Assay Reference Genome
          Location

          Chr.20:57224291 on build GRCh38
          Cytoband
          20q13.31
          Assay ID Hs07207836_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          655

          Gene Name:

          bone morphogenetic protein 7

          Gene Aliases:

          OP-1

          Location:

          Chr.20:57168753-57266651 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          BMP7 NM_001719.2 NP_001710.1
          AK094784.1
          AK291186.1
          AK312419.1
          BC008584.1 AAH08584.1
          EU679366.1
          M60316.1 AAA36738.1
          X51801.1 CAA36100.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3646210 Chr.20:57220830 - 57226489 on Build GRCh38 Loss BMP7

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs144325716] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          ossification
          metanephros development
          ureteric bud development
          mesoderm formation
          mesonephros development
          epithelial to mesenchymal transition
          axon guidance
          embryonic pattern specification
          negative regulation of striated muscle cell apoptotic process
          positive regulation of peptidyl-threonine phosphorylation
          positive regulation of pathway-restricted SMAD protein phosphorylation
          dendrite development
          embryonic limb morphogenesis
          positive regulation of bone mineralization
          BMP signaling pathway
          epithelial cell differentiation
          negative regulation of NF-kappaB transcription factor activity
          response to estradiol
          response to vitamin D
          positive regulation of heterotypic cell-cell adhesion
          protein localization to nucleus
          growth
          negative regulation of phosphorylation
          negative regulation of NF-kappaB import into nucleus
          odontogenesis of dentin-containing tooth
          regulation of apoptotic process
          positive regulation of apoptotic process
          steroid hormone mediated signaling pathway
          negative regulation of MAP kinase activity
          response to peptide hormone
          negative regulation of neuron differentiation
          positive regulation of osteoblast differentiation
          negative regulation of Notch signaling pathway
          negative regulation of cell cycle
          negative regulation of mitotic nuclear division
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          embryonic camera-type eye morphogenesis
          branching morphogenesis of an epithelial tube
          mesenchymal cell differentiation
          neuron projection morphogenesis
          cartilage development
          embryonic skeletal joint morphogenesis
          regulation of pathway-restricted SMAD protein phosphorylation
          SMAD protein signal transduction
          branching involved in salivary gland morphogenesis
          mesenchyme development
          negative regulation of cell death
          negative regulation of prostatic bud formation
          regulation of branching involved in prostate gland morphogenesis
          monocyte aggregation
          cellular response to hypoxia
          cellular response to BMP stimulus
          negative regulation of mesenchymal cell apoptotic process involved in nephron morphogenesis
          negative regulation of glomerular mesangial cell proliferation
          metanephric mesenchyme morphogenesis
          nephrogenic mesenchyme morphogenesis
          metanephric mesenchymal cell proliferation involved in metanephros development
          positive regulation of dendrite development
          positive regulation of hyaluranon cable assembly
          regulation of removal of superoxide radicals
          cytokine activity
          transforming growth factor beta receptor binding
          protein binding
          growth factor activity
          heparin binding
          BMP receptor binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline