Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07226371_cn
          See other AP4B1-AS1 CNV Assays ›
          Gene Symbol
          AP4B1-AS1
          Assay Reference Genome
          Location

          Chr.1:113852427 on build GRCh38
          Cytoband
          Assay ID Hs07226371_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          100287722

          Gene Name:

          AP4B1 antisense RNA 1

          Gene Aliases:

          -

          Location:

          Chr.1:113812612-113901237 on Build GRCh38

          Assay Gene Location:

          Within Intron 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          AP4B1-AS1 NR_125965.1
          DB028504.1
          DB074478.1

          Target Gene Details

          Entrez Gene ID:

          26191

          Gene Name:

          protein tyrosine phosphatase non-receptor type 22

          Gene Aliases:

          LYP, LYP1, LYP2, PEP, PTPN22.5, PTPN22.6, PTPN8

          Location:

          Chr.1:113813811-113871759 on Build GRCh38

          Assay Gene Location:

          Within Intron 10
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PTPN22 NM_001193431.3 NP_001180360.2
          NM_001308297.2 NP_001295226.2
          NM_012411.6 NP_036543.5
          NM_015967.8 NP_057051.4
          XM_011541221.2 XP_011539523.1
          XM_011541222.2 XP_011539524.1
          XM_011541223.3 XP_011539525.1
          XM_011541225.3 XP_011539527.1
          XM_017001005.3 XP_016856494.1
          XM_017001006.2 XP_016856495.1
          XM_047417630.1 XP_047273586.1
          XM_047417631.1 XP_047273587.1
          XM_047417632.1 XP_047273588.1
          AF001846.1 AAD00904.1
          AF077031.1 AAD27764.1
          AF150732.1 AAF67472.1
          AK303124.1 BAG64230.1
          AK310570.1
          AK310698.1
          BC017785.1 AAH17785.1
          BC071670.1 AAH71670.1
          BK008060.1
          BK008061.1
          GU479452.1 ADD59979.1
          JN084012.1 AEL79472.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3587240 Chr.1:113533246 - 114158910 on Build GRCh38 Loss PTPN22 PHTF1 AP4B1-AS1 BCL2L15 HIPK1 DCLRE1B SYT6 OLFML3 RSBN1 MAGI3 HIPK1-AS1 AP4B1
          esv3587239 Chr.1:113510852 - 113866662 on Build GRCh38 Loss PTPN22 PHTF1 AP4B1-AS1 RSBN1 MAGI3
          nsv5425010 Chr.1:113852295 - 113853484 on Build GRCh38 Deletion PTPN22 AP4B1-AS1
          esv3692857 Chr.1:112712767 - 113894764 on Build GRCh38 Loss PPM1J-DT SLC16A1-AS1 NUTF2P4 LRIG2-DT AKR7A2P1 RSBN1 MIR11399 SLC16A1 PTPN22 PHTF1 AP4B1-AS1 BCL2L15 LRIG2 LINC01356 LOC643441 PPM1J MAGI3 TAFA3 LINC01357 AP4B1
          esv23869 Chr.1:113320330 - 114358495 on Build GRCh38 Loss PTPN22 PHTF1 AP4B1-AS1 BCL2L15 HIPK1 DCLRE1B SYT6 OLFML3 RSBN1 MAGI3 HIPK1-AS1 AP4B1
          esv3587250 Chr.1:113852261 - 113853452 on Build GRCh38 Loss PTPN22 AP4B1-AS1

          Back To Top

          More Information


          Additional Information:

          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation Validated

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          lipid metabolic process
          autophagy
          negative regulation of autophagy
          positive regulation of gene expression
          negative regulation of gene expression
          T cell differentiation
          lipopolysaccharide-mediated signaling pathway
          positive regulation of type I interferon production
          response to lipopolysaccharide
          negative regulation of interleukin-6 production
          negative regulation of interleukin-8 production
          negative regulation of tumor necrosis factor production
          positive regulation of interferon-gamma production
          regulation of natural killer cell proliferation
          positive regulation of toll-like receptor 3 signaling pathway
          positive regulation of toll-like receptor 4 signaling pathway
          phosphoanandamide dephosphorylation
          negative regulation of JUN kinase activity
          regulation of innate immune response
          T cell receptor signaling pathway
          regulation of B cell receptor signaling pathway
          regulation of T cell receptor signaling pathway
          negative regulation of T cell receptor signaling pathway
          negative regulation of T cell activation
          positive regulation of ERK1 and ERK2 cascade
          negative regulation of nucleotide-binding oligomerization domain containing 2 signaling pathway
          cellular response to muramyl dipeptide
          positive regulation of NLRP3 inflammasome complex assembly
          regulation of NIK/NF-kappaB signaling
          positive regulation of protein K63-linked ubiquitination
          negative regulation of p38MAPK cascade
          phosphoprotein phosphatase activity
          protein tyrosine phosphatase activity
          non-membrane spanning protein tyrosine phosphatase activity
          protein binding
          hydrolase activity
          phosphatase activity
          SH3 domain binding
          kinase binding
          ubiquitin protein ligase binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline