Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07438154_cn
          See other PFKP CNV Assays ›
          Gene Symbol
          PFKP
          Assay Reference Genome
          Location

          Chr.10:3126192 on build GRCh38
          Cytoband
          Assay ID Hs07438154_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5214

          Gene Name:

          phosphofructokinase, platelet

          Gene Aliases:

          ATP-PFK, PFK-C, PFK-P, PFKF

          Location:

          Chr.10:3067548-3136805 on Build GRCh38

          Assay Gene Location:

          Within Intron 21
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PFKP NM_001242339.2 NP_001229268.1
          NM_001323067.2 NP_001309996.1
          NM_001323068.2 NP_001309997.1
          NM_001323069.2 NP_001309998.1
          NM_001323070.1 NP_001309999.1
          NM_001323071.2 NP_001310000.1
          NM_001323072.2 NP_001310001.1
          NM_001323073.1 NP_001310002.1
          NM_001323074.1 NP_001310003.1
          NM_001345944.1 NP_001332873.1
          NM_001410880.1 NP_001397809.1
          NM_002627.5 NP_002618.1
          XM_005252466.5 XP_005252523.1
          XM_006717449.2 XP_006717512.1
          XM_047425350.1 XP_047281306.1
          AK092597.1 BAG52577.1
          AK126153.1 1 858
          AK128373.1
          AK130767.1 1 197
          AK291841.1 BAF84530.1
          AK308003.1
          AK308226.1
          AK309692.1
          BC002536.1 AAH02536.1
          BC021698.1 AAH21698.1
          BC029138.1 AAH29138.1
          D21863.1 BAA21891.1
          M64784.1 AAA36435.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv549814 Chr.10:3055020 - 3170233 on Build GRCh38 Loss SNORD142 PITRM1-AS1 LOC124902536 PITRM1 PFKP-DT PFKP
          dgv932n54 Chr.10:2382612 - 3253481 on Build GRCh38 Gain SNORD142 PITRM1-AS1 LOC107983989 LOC105376350 LOC124902536 PITRM1 LINC02668 LOC105376353 PFKP-DT LINC02645 PFKP
          nsv428229 Chr.10:2618610 - 3156581 on Build GRCh38 Gain SNORD142 PITRM1-AS1 LOC105376350 LOC124902536 PITRM1 PFKP-DT PFKP
          esv2730995 Chr.10:2770220 - 3706473 on Build GRCh38 Deletion SNORD142 PITRM1-AS1 LOC124902536 PITRM1 LOC105376360 LINC02668 LINC02669 LOC105376353 PFKP-DT PFKP LOC124902565
          nsv6442021 Chr.10:3030896 - 3173781 on Build GRCh38 Duplication SNORD142 PITRM1-AS1 LOC124902536 PITRM1 PFKP-DT PFKP
          esv2750891 Chr.10:3098894 - 3164173 on Build GRCh38 Loss SNORD142 PITRM1-AS1 PITRM1 PFKP
          nsv6442625 Chr.10:3121305 - 3126563 on Build GRCh38 Deletion PFKP
          nsv6442520 Chr.10:2846098 - 3581354 on Build GRCh38 Duplication SNORD142 PITRM1-AS1 LOC124902536 PITRM1 LOC105376360 LINC02668 LINC02669 LOC105376353 PFKP-DT PFKP LOC124902565
          nsv549810 Chr.10:3035272 - 3691194 on Build GRCh38 Gain SNORD142 PITRM1-AS1 LOC124902536 PITRM1 LOC105376360 LINC02668 LINC02669 LOC105376353 PFKP-DT PFKP LOC124902565
          nsv6454219 Chr.10:2905224 - 3284256 on Build GRCh38 Duplication SNORD142 PITRM1-AS1 LOC124902536 PITRM1 LINC02668 LOC105376353 PFKP-DT PFKP
          nsv6437272 Chr.10:3058401 - 3259500 on Build GRCh38 Deletion SNORD142 PITRM1-AS1 LOC124902536 PITRM1 LINC02668 LOC105376353 PFKP-DT PFKP
          nsv549818 Chr.10:3081241 - 3227125 on Build GRCh38 Gain SNORD142 PITRM1-AS1 PITRM1 PFKP
          nsv525229 Chr.10:3123124 - 3127579 on Build GRCh38 Loss PFKP
          nsv4179484 Chr.10:2460844 - 3257750 on Build GRCh38 Duplication SNORD142 PITRM1-AS1 LOC107983989 LOC105376350 LOC124902536 PITRM1 LINC02668 LOC105376353 PFKP-DT LINC02645 PFKP
          dgv38n206 Chr.10:2613782 - 3185857 on Build GRCh38 Duplication SNORD142 PITRM1-AS1 LOC105376350 LOC124902536 PITRM1 PFKP-DT PFKP
          nsv6455282 Chr.10:3085901 - 3154200 on Build GRCh38 Deletion SNORD142 PITRM1-AS1 PITRM1 PFKP

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          fructose 6-phosphate metabolic process
          glycolytic process
          fructose 1,6-bisphosphate metabolic process
          canonical glycolysis
          cellular response to leukemia inhibitory factor
          nucleotide binding
          catalytic activity
          6-phosphofructokinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          identical protein binding
          macromolecular complex binding
          cadherin binding
          metal ion binding
          fructose-6-phosphate binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline