Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07515680_cn
          See other SMIM9 CNV Assays ›
          Gene Symbol
          SMIM9
          Assay Reference Genome
          Location

          Chr.X:154826852 on build GRCh38
          Cytoband
          Assay ID Hs07515680_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          100132963

          Gene Name:

          small integral membrane protein 9

          Gene Aliases:

          CXorf68

          Location:

          Chr.X:154823348-154834662 on Build GRCh38

          Assay Gene Location:

          Within Intron 4
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SMIM9 NM_001162936.4 NP_001156408.1
          AW072401.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv33784 Chr.X:153962665 - 155699588 on Build GRCh38 Gain+Loss FAM50A OPN1MW2 MIR718 G6PD F8A1 DNASE1L1 SNORA36A GAB3 ATP6AP1-DT FAM223B OPN1MW3 MTCP1 SPRY3 ATP6AP1 HCFC1 MIR6858 MIR1184-2 SNORA56 GDI1 TEX28 TMLHE SLC10A3 IKBKG MIR1184-3 LOC101927830 EMD FAM223A HCFC1-AS1 RAB39B F8 CMC4 LAGE3 MPP1 MECP2 TMEM187 F8A3 BRCC3 H2AB2 H2AB3 CLIC2 CTAG1B TAFAZZIN SMIM9 CTAG2 FLNA FAM3A UBL4A DKC1 TMLHE-AS1 OPN1LW PLXNA3 F8A2 MIR3202-2 MIR664B TKTL1 H2AB1 OPN1MW IRAK1 MIR1184-1 RPL10 VBP1 FUNDC2 SNORA70 MIR3202-1 LOC105373386 CTAG1A
          nsv5878993 Chr.X:154598638 - 155803675 on Build GRCh38 Deletion F8A3 BRCC3 F8A1 H2AB2 SNORA36A GAB3 H2AB3 FAM223B CLIC2 CTAG1B MTCP1 SMIM9 CTAG2 SPRY3 DKC1 TMLHE-AS1 MIR1184-2 SNORA56 F8A2 TMLHE MIR664B H2AB1 MIR1184-3 LOC101927830 MIR1184-1 VBP1 RAB39B FUNDC2 F8 CMC4 MPP1
          nsv516158 Chr.X:154790808 - 154831402 on Build GRCh38 Gain SMIM9 MPP1
          esv3577568 Chr.X:154820215 - 154899365 on Build GRCh38 Gain MIR1184-1 SMIM9 F8 F8A1 H2AB1
          esv29972 Chr.X:154781072 - 155699751 on Build GRCh38 Gain MIR1184-2 F8A3 BRCC3 F8A2 F8A1 TMLHE H2AB2 H2AB1 MIR1184-3 H2AB3 LOC101927830 CLIC2 MIR1184-1 MTCP1 VBP1 RAB39B FUNDC2 SMIM9 F8 SPRY3 CMC4 MPP1 TMLHE-AS1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline