Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07535236_cn
          See other CRIP2 CNV Assays ›
          Gene Symbol
          CRIP2
          Assay Reference Genome
          Location

          Chr.14:105475116 on build GRCh38
          Cytoband
          14q32.33
          Assay ID Hs07535236_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          1397

          Gene Name:

          cysteine rich protein 2

          Gene Aliases:

          CRIP, CRP2, ESP1

          Location:

          Chr.14:105472938-105480170 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CRIP2 NM_001270837.1 NP_001257766.1
          NM_001270841.1 NP_001257770.1
          NM_001312.3 NP_001303.1
          NR_073081.1
          NR_073082.1
          NR_073083.1
          AK057837.1
          AK223251.1 BAD96971.1
          AK300092.1
          AK315757.1
          BC000434.2 AAH00434.1
          BC001931.1 AAH01931.1
          BC034151.1 AAH34151.1
          BC128101.1
          BE271979.1
          BT019911.1 AAV38714.1
          CN367966.1
          D42123.1 BAA07703.1
          HY078632.1
          HY161884.1
          U36190.1 AAB03194.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv566189 Chr.14:105305091 - 105475240 on Build GRCh38 Gain PACS2 BRF1 LOC100507437 MTA1 CRIP2 TEX22
          dgv3994n54 Chr.14:105474172 - 105477662 on Build GRCh38 Gain CRIP2
          nsv566259 Chr.14:105474444 - 105477342 on Build GRCh38 Gain+Loss CRIP2
          nsv827088 Chr.14:105454469 - 105498549 on Build GRCh38 Gain CRIP1 C14orf80 MTA1 CRIP2
          nsv952282 Chr.14:105363764 - 105510163 on Build GRCh38 Deletion PACS2 CRIP1 LOC100507437 C14orf80 MTA1 CRIP2 TEX22
          dgv3997n54 Chr.14:105474383 - 105475240 on Build GRCh38 Loss CRIP2
          nsv566265 Chr.14:105474766 - 105479048 on Build GRCh38 Loss CRIP2
          nsv1054190 Chr.14:105246882 - 105701138 on Build GRCh38 Gain IGHA2 IGHG2 CRIP1 LOC100507437 MIR8071-1 CRIP2 ELK2AP IGHG4 TMEM121 LOC105370697 PACS2 BRF1 MIR8071-2 C14orf80 MTA1 IGH BTBD6 TEX22
          nsv517055 Chr.14:105136282 - 106779068 on Build GRCh38 Gain+Loss IGHV3-48 IGHA2 IGHG2 MIR4537 IGHV4-55 NUDT14 CRIP2 IGHA1 IGHV1-69 IGHG4 TMEM121 PACS2 BRF1 IGHV3-69-1 MIR8071-2 IGHV3-23 LINC00226 IGHV3-71 IGHV4-31 C14orf80 IGHG1 MIR4507 IGHM KIAA0125 LOC107987211 BTBD6 TEX22 IGHV3-41 ADAM6 IGHD CRIP1 LOC100507437 MIR4539 IGHV3-30 MIR8071-1 ELK2AP MIR4538 LOC105370697 MIR6765 JAG2 MTA1 IGH IGHG3 LINC00221
          nsv566220 Chr.14:105472502 - 105475240 on Build GRCh38 Gain+Loss CRIP2
          dgv3996n54 Chr.14:105474226 - 105488519 on Build GRCh38 Gain CRIP1 CRIP2
          nsv566204 Chr.14:105376842 - 105479048 on Build GRCh38 Gain PACS2 LOC100507437 MTA1 CRIP2 TEX22
          dgv3998n54 Chr.14:105474495 - 105770768 on Build GRCh38 Loss IGHA2 IGHG2 CRIP1 MIR8071-1 CRIP2 ELK2AP IGHA1 IGHG4 TMEM121 LOC105370697 MIR8071-2 C14orf80 IGHG1 IGH IGHG3 LOC107987211
          nsv817647 Chr.14:105382554 - 105480505 on Build GRCh38 Gain PACS2 LOC100507437 MTA1 CRIP2 TEX22
          nsv1160204 Chr.14:105291411 - 105483783 on Build GRCh38 Duplication PACS2 BRF1 LOC100507437 MTA1 CRIP2 TEX22
          esv21791 Chr.14:105474144 - 105475279 on Build GRCh38 Loss CRIP2
          nsv566206 Chr.14:105439736 - 105543235 on Build GRCh38 Loss TMEM121 LOC105370697 CRIP1 C14orf80 MTA1 CRIP2
          nsv832890 Chr.14:105304955 - 105483089 on Build GRCh38 Loss PACS2 BRF1 LOC100507437 MTA1 CRIP2 TEX22
          nsv1440 Chr.14:105473170 - 105479862 on Build GRCh38 Insertion CRIP2
          dgv3989n54 Chr.14:105468704 - 105475240 on Build GRCh38 Gain MTA1 CRIP2
          nsv566209 Chr.14:105465248 - 105475240 on Build GRCh38 Gain+Loss MTA1 CRIP2
          dgv3999n54 Chr.14:105474867 - 105487998 on Build GRCh38 Loss CRIP1 CRIP2
          esv2760014 Chr.14:105047009 - 105483089 on Build GRCh38 Loss PACS2 MIR6765 BRF1 GPR132 JAG2 LOC100507437 MTA1 NUDT14 CRIP2 LOC102723354 BTBD6 TEX22
          dgv3988n54 Chr.14:105466757 - 105477662 on Build GRCh38 Gain MTA1 CRIP2
          nsv470679 Chr.14:105124547 - 105605978 on Build GRCh38 Loss IGHA2 CRIP1 LOC100507437 NUDT14 CRIP2 TMEM121 LOC105370697 PACS2 MIR6765 BRF1 JAG2 C14orf80 MTA1 IGH BTBD6 TEX22
          dgv3987n54 Chr.14:105461599 - 105499970 on Build GRCh38 Loss CRIP1 C14orf80 MTA1 CRIP2
          esv3635645 Chr.14:105473682 - 105484540 on Build GRCh38 Gain CRIP2

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell proliferation
          hemopoiesis
          zinc ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline