Search
Search

Entrez Gene ID: | 67196 |
Gene Name: | ubiquitin-conjugating enzyme E2T (putative) |
Gene Aliases: |
2700084L22Rik, C80607 |
Location: |
Chr.1:134962565-134974180 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2t | NM_001278115.1 | NP_001265044.1 | ||
| NM_026024.3 | NP_080300.1 | |||
| AK012565.1 | BAB28320.1 | |||
| AK021213.1 | BAB32332.1 | |||
| AK166098.1 | ||||
| BC029213.1 | AAH29213.1 | |||
| CK334733.1 |
Set Membership: |
|