Search
Search

Entrez Gene ID: | 12841 |
Gene Name: | collagen, type IX, alpha 3 |
Gene Aliases: |
AV006866 |
Location: |
Chr.2:180597795-180622185 on Build GRCm38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Col9a3 | NM_009936.2 | NP_034066.2 | ||
| AF349718.1 | AAL56219.1 | |||
| AK030350.1 | BAC26915.1 | |||
| AK038481.1 |