Search
Search

Entrez Gene ID: | 16561 |
Gene Name: | kinesin family member 1B |
Gene Aliases: |
A530096N05Rik, AI448212, AI506502, D4Mil1e, KIF1Bp130, KIF1Bp204 |
Location: |
Chr.4:149176319-149307733 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 44 - Exon 44 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kif1b | NM_001290995.1 | NP_001277924.1 | ||
| NM_207682.2 | NP_997565.2 | |||
| AB023656.1 | BAA75243.1 | |||
| AF090190.1 | AAF06718.1 | |||
| AF131865.1 | AAD39438.1 | |||
| AK147409.1 | ||||
| CF746574.1 |
Set Membership: |
|