Search
Search

Entrez Gene ID: | 233789 |
Gene Name: | SMG1 homolog, phosphatidylinositol 3-kinase-related kinase (C. elegans) |
Gene Aliases: |
2610207I05Rik, 5430435M13Rik, C130002K18Rik, mKIAA0421 |
Location: |
Chr.7:118131308-118243658 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smg1 | NM_001031814.1 | NP_001026984.1 | ||
| AK041264.1 | BAC30884.2 | |||
| AK157798.1 |
Set Membership: |
|