Search
Search

Entrez Gene ID: | 68337 |
Gene Name: | cysteine rich protein 2 |
Gene Aliases: |
0610010I23Rik, AW743261, C77570, CRP2, CRP4, Crp, ESP1, Hlp |
Location: |
Chr.12:113140236-113145506 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crip2 | NM_024223.2 | NP_077185.1 | ||
| AF469648.1 | AAM89218.1 | |||
| AK002484.1 | BAB22136.1 | |||
| BC002093.1 | AAH02093.1 | |||
| BC002096.1 | AAH02096.1 | |||
| CT010309.1 | CAJ18517.1 | |||
| CX217539.1 |
Set Membership: |
|