Search
Search

Entrez Gene ID: | 20103 |
Gene Name: | ribosomal protein S5 |
Gene Aliases: |
AA617411 |
Location: |
Chr.7:12922311-12926686 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps5 | NM_009095.2 | NP_033121.2 | ||
| AK002235.1 | BAB21953.1 | |||
| AK009663.1 | BAB26424.1 | |||
| AK010681.1 | BAB27113.1 | |||
| AK012424.1 | BAB28229.1 | |||
| AK012481.1 | BAB28270.1 | |||
| AK020477.1 | BAB32115.1 | |||
| AK020754.1 | BAB32203.1 | |||
| AK050611.1 | BAC34342.1 | |||
| AK050622.1 | BAC34347.1 | |||
| AK152449.1 | BAE31228.1 | |||
| AK160552.1 | BAE35868.1 | |||
| AK164079.1 | BAE37616.1 | |||
| AK167533.1 | BAE39602.1 | |||
| AK168044.1 | BAE40026.1 | |||
| AK168106.1 | BAE40077.1 | |||
| AK169016.1 | BAE40813.1 | |||
| AK172446.1 | BAE43008.1 | |||
| AK191975.1 | ||||
| BC058690.1 | AAH58690.1 | |||
| U78085.1 | AAB63526.1 | |||
| Y12431.1 | CAA73041.1 |
Set Membership: |
|