Search
Search

Entrez Gene ID: | 18788 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 2 |
Gene Aliases: |
PAI-2, Planh2, ovalbumin |
Location: |
Chr.1:107500892-107525600 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb2 | NM_001174170.1 | NP_001167641.1 | ||
| NM_011111.4 | NP_035241.1 | |||
| AK081487.1 | BAC38231.1 | |||
| AK090049.1 | BAC41065.1 | |||
| BC138815.1 | ||||
| BC138817.1 | ||||
| X16490.1 | CAA34507.1 |
Set Membership: |
|