Search
Search

Entrez Gene ID: | 12009 |
Gene Name: | centrosomal protein 131 |
Gene Aliases: |
AZ1, Azi, Azi1 |
Location: |
Chr.11:120064429-120086859 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep131 | NM_009734.3 | NP_033864.3 | ||
| AK133245.1 | ||||
| BC141108.1 | ||||
| BY724850.1 | ||||
| D43921.1 | BAA07868.1 |
Set Membership: |
|