Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00149817_cn
          See other KRTAP10-3 CNV Assays ›
          Gene Symbol
          KRTAP10-3
          Assay Reference Genome
          Location

          Chr.21:44558028 on build GRCh38
          Cytoband
          Assay ID Hs00149817_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          386682

          Gene Name:

          keratin associated protein 10-3

          Gene Aliases:

          KAP10.3, KAP18-3, KAP18.3, KRTAP10.3, KRTAP18-3, KRTAP18.3

          Location:

          Chr.21:44557790-44558795 on Build GRCh38

          Assay Gene Location:

          Within Exon 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          KRTAP10-3 NM_198696.3 1 768 NP_941969.2
          AB076350.1 BAD01537.1
          AJ566383.1 1 192 CAD97463.1
          BC133677.1 AAI33678.1

          Target Gene Details

          Entrez Gene ID:

          54084

          Gene Name:

          thrombospondin type laminin G domain and EAR repeats

          Gene Aliases:

          C21orf29, DFNB98, ECTD14, STHAG10, TSP-EAR

          Location:

          Chr.21:44497893-44711572 on Build GRCh38

          Assay Gene Location:

          Within Intron 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          TSPEAR NM_001272037.2 NP_001258966.1
          NM_144991.3 NP_659428.2
          AJ487962.1 CAD32309.1
          BC021197.2 AAH21197.2
          BC115415.1 AAI15416.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6540525 Chr.21:44521952 - 44596004 on Build GRCh38 Duplication KRTAP10-6 KRTAP10-1 KRTAP10-5 KRTAP10-2 TSPEAR KRTAP10-4 KRTAP10-3
          nsv587803 Chr.21:44514317 - 44569605 on Build GRCh38 Loss KRTAP10-1 TSPEAR-AS2 KRTAP10-2 TSPEAR KRTAP10-3 TSPEAR-AS1
          esv3647132 Chr.21:44539529 - 44591271 on Build GRCh38 Gain KRTAP10-6 KRTAP10-1 KRTAP10-5 KRTAP10-2 TSPEAR KRTAP10-4 KRTAP10-3
          esv2723638 Chr.21:44249174 - 44593709 on Build GRCh38 Deletion KRTAP10-6 KRTAP10-1 CFAP410 DNMT3L KRTAP10-2 TSPEAR KRTAP10-4 KRTAP10-3 TSPEAR-AS1 PFKL DNMT3L-AS1 KRTAP10-5 TRPM2-AS TRPM2 LRRC3-DT TSPEAR-AS2 AIRE LRRC3 LINC02575
          nsv4288498 Chr.21:44551317 - 44558317 on Build GRCh38 Deletion KRTAP10-2 TSPEAR KRTAP10-3
          nsv6535810 Chr.21:44473788 - 44670842 on Build GRCh38 Duplication KRTAP10-6 KRTAP10-1 KRTAP12-3 KRTAP12-2 KRTAP10-11 KRTAP10-2 KRTAP10-9 TSPEAR KRTAP10-7 KRTAP10-4 KRTAP10-10 KRTAP10-3 TSPEAR-AS1 KRTAP10-5 KRTAP10-8 TSPEAR-AS2 KRTAP12-4 LINC02575
          nsv1055163 Chr.21:44475780 - 44646211 on Build GRCh38 Gain KRTAP10-6 KRTAP10-1 KRTAP10-2 KRTAP10-9 TSPEAR KRTAP10-7 KRTAP10-4 KRTAP10-10 KRTAP10-3 TSPEAR-AS1 KRTAP10-5 KRTAP10-8 TSPEAR-AS2 LINC02575
          nsv6546352 Chr.21:44332501 - 44603700 on Build GRCh38 Deletion KRTAP10-6 KRTAP10-1 CFAP410 KRTAP10-2 TSPEAR KRTAP10-7 KRTAP10-4 KRTAP10-3 TSPEAR-AS1 KRTAP10-5 TRPM2-AS TRPM2 LRRC3-DT TSPEAR-AS2 LRRC3 LINC02575
          esv33817 Chr.21:43749531 - 44647625 on Build GRCh38 Loss KRTAP10-6 CSTB KRTAP10-1 CFAP410 DNMT3L RRP1 KRTAP10-11 KRTAP10-2 AATBC KRTAP10-7 KRTAP10-10 KRTAP10-3 TSPEAR-AS1 PFKL DNMT3L-AS1 TRAPPC10 KRTAP10-5 TRPM2 LRRC3 LINC02575 LINC01678 ICOSLG AGPAT3 PWP2 KRTAP10-9 TSPEAR KRTAP10-4 PDXK TRPM2-AS LRRC3-DT KRTAP10-8 TSPEAR-AS2 GATD3 AIRE LOC105372832
          nsv4283103 Chr.21:43950580 - 44658488 on Build GRCh38 Duplication KRTAP10-6 KRTAP10-1 CFAP410 DNMT3L KRTAP10-11 KRTAP10-2 KRTAP10-7 KRTAP10-10 KRTAP10-3 TSPEAR-AS1 PFKL DNMT3L-AS1 TRAPPC10 KRTAP10-5 TRPM2 KRTAP12-4 LRRC3 LINC02575 LINC01678 ICOSLG KRTAP12-3 AGPAT3 PWP2 KRTAP10-9 TSPEAR KRTAP10-4 TRPM2-AS LRRC3-DT KRTAP10-8 TSPEAR-AS2 GATD3 AIRE LOC105372832
          dgv4435n100 Chr.21:44527151 - 44595409 on Build GRCh38 Gain KRTAP10-6 KRTAP10-1 KRTAP10-5 KRTAP10-2 TSPEAR KRTAP10-4 KRTAP10-3

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s)

          signal transduction
          Notch signaling pathway
          sensory perception of sound
          regulation of Notch signaling pathway
          hair cycle process
          tooth mineralization

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline