Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00158829_cn
          See other NAPA CNV Assays ›
          Gene Symbol
          NAPA
          Assay Reference Genome
          Location

          Chr.19:47492074 on build GRCh38
          Cytoband
          Assay ID Hs00158829_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          8775

          Gene Name:

          NSF attachment protein alpha

          Gene Aliases:

          SNAPA

          Location:

          Chr.19:47484530-47515063 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 8 - Exon 9
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          NAPA NM_003827.4 NP_003818.2
          NR_038456.2
          NR_038457.2
          XM_011527436.2 XP_011525738.1
          AK098401.1
          AK126519.1 2 2967
          AK292244.1 BAF84933.1
          BC001165.1 AAH01165.1
          BC007432.2 AAH07432.1
          BC028234.1 AAH28234.1
          BC091511.1 AAH91511.1
          BT019568.1 AAV38375.1
          BU168063.1
          CX873074.1
          U39412.1 AAC80170.1

          Target Gene Details

          Entrez Gene ID:

          100505681

          Gene Name:

          NAPA antisense RNA 1

          Gene Aliases:

          -

          Location:

          Chr.19:47484282-47501597 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          NAPA-AS1 NR_038452.1
          AK057858.1
          BG035979.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv428366 Chr.19:47354499 - 47522248 on Build GRCh38 Loss DHX34 ZNF541 MEIS3 NAPA SLC8A2 KPTN NAPA-AS1
          nsv1056887 Chr.19:47481963 - 47527369 on Build GRCh38 Gain ZNF541 NAPA KPTN NAPA-AS1
          nsv579888 Chr.19:47428355 - 47658874 on Build GRCh38 Gain ZNF541 BICRA-AS2 NAPA SLC8A2 BICRA KPTN NAPA-AS1
          nsv953591 Chr.19:47486744 - 47500943 on Build GRCh38 Deletion NAPA NAPA-AS1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          intra-Golgi vesicle-mediated transport
          brain development
          regulation of synaptic vesicle priming
          protein transport
          synaptic vesicle priming
          vesicle-mediated transport
          neuron differentiation
          protein-containing complex disassembly
          synaptic transmission, glutamatergic
          SNARE complex disassembly
          apical protein localization
          membrane fusion
          SNARE binding
          soluble NSF attachment protein activity
          protein binding
          syntaxin binding
          macromolecular complex binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline