Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00175239_cn
          See other ZIC5 CNV Assays ›
          Gene Symbol
          ZIC5
          Assay Reference Genome
          Location

          Chr.13:99971544 on build GRCh38
          Cytoband
          13q32.3
          Assay ID Hs00175239_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          85416

          Gene Name:

          Zic family member 5

          Gene Aliases:

          -

          Location:

          Chr.13:99963021-99971924 on Build GRCh38

          Assay Gene Location:

          Within Exon 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          ZIC5 NM_033132.3 1 381 NP_149123.2
          AF378304.1 1 366 AAK55418.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv832697 Chr.13:99936254 - 100109792 on Build GRCh38 Loss LOC101927437 LOC105370333 ZIC5 ZIC2 PCCA LINC00554
          dgv347n67 Chr.13:99953652 - 100005016 on Build GRCh38 Gain LOC101927437 ZIC5 ZIC2 LINC00554
          dgv346n67 Chr.13:99948584 - 100022042 on Build GRCh38 Gain LOC101927437 ZIC5 ZIC2 LINC00554
          esv2747875 Chr.13:99669768 - 100007232 on Build GRCh38 Deletion CLYBL-AS1 LOC101927437 CLYBL CLYBL-AS2 ZIC5 ZIC2 LINC00554
          nsv826757 Chr.13:99926725 - 100028029 on Build GRCh38 Loss LOC101927437 ZIC5 ZIC2 LINC00554
          nsv1118272 Chr.13:99969546 - 99971746 on Build GRCh38 Deletion ZIC5
          nsv456093 Chr.13:99965051 - 100014379 on Build GRCh38 Loss ZIC5 ZIC2 LINC00554
          nsv826766 Chr.13:99961399 - 99995462 on Build GRCh38 Gain ZIC5 ZIC2 LINC00554
          nsv1069768 Chr.13:99970245 - 99971646 on Build GRCh38 Deletion ZIC5
          nsv562952 Chr.13:99934267 - 99976973 on Build GRCh38 Loss LOC101927437 ZIC5
          nsv510622 Chr.13:99942336 - 100039611 on Build GRCh38 Deletion LOC101927437 ZIC5 ZIC2 LINC00554
          dgv345n67 Chr.13:99908442 - 100033389 on Build GRCh38 Gain LOC101927437 CLYBL ZIC5 ZIC2 LINC00554
          nsv562954 Chr.13:99967208 - 99986034 on Build GRCh38 Loss ZIC5 ZIC2

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          nervous system development
          cell differentiation
          DNA binding
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lq2bv:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0