Search
Search

Entrez Gene ID: | 54961 |
Gene Name: | slingshot protein phosphatase 3 |
Gene Aliases: |
SSH3L |
Location: |
Chr.11:67303448-67312607 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSH3 | NM_017857.3 | 14 | 2682 | NP_060327.3 |
| XM_017017942.1 | 14 | 2721 | XP_016873431.1 | |
| AB072360.3 | 14 | 2406 | BAB84119.3 | |
| AF085851.1 | 1 | 386 | ||
| AK000522.1 | 10 | 1855 | BAA91228.1 | |
| AK001790.1 | 8 | 1567 | BAA91913.1 | |
| AK074432.1 | 14 | 2600 | BAB85080.1 | |
| AK094226.1 | 13 | 2711 | BAC04314.1 | |
| AK124234.1 | 1 | 1706 | ||
| BC007709.1 | 14 | 2561 | AAH07709.1 | |
| BU689839.1 | 1 | 213 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555253 | Chr.11:67265605 - 67397024 on Build GRCh38 | Loss |
|
Set Membership: |
|