Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00233222_cn
          See other MCL1 CNV Assays ›
          Gene Symbol
          MCL1
          Assay Reference Genome
          Location

          Chr.1:150579065 on build GRCh38
          Cytoband
          Assay ID Hs00233222_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          4170

          Gene Name:

          MCL1 apoptosis regulator, BCL2 family member

          Gene Aliases:

          BCL2L3, EAT, MCL1-ES, MCL1L, MCL1S, Mcl-1, TM, bcl2-L-3, mcl1/EAT

          Location:

          Chr.1:150574558-150579610 on Build GRCh38

          Assay Gene Location:

          Within Exon 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          MCL1 NM_001197320.2 NP_001184249.1
          NM_021960.5 1 546 NP_068779.1
          NM_182763.3 1 546 NP_877495.1
          AF118124.1 1 515 AAD13299.1
          AF203373.1 1 466 AAG00904.1
          AK294462.1 2 246 BAG57694.1
          AK297217.1 BAG59700.1
          AK300499.1 2 320 BAG62213.1
          AK304775.1 2 356 BAG65530.1
          AK312508.1 1 546 BAG35409.1
          AK316267.1 2 502 BAH14638.1
          BC017197.2 1 520 AAH17197.1
          BC071897.1 1 522 AAH71897.1
          BC107735.1 1 537 AAI07736.1
          BT006640.1 1 466 AAP35286.1
          L08246.1 1 526

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1006838 Chr.1:150453465 - 150750307 on Build GRCh38 Gain ECM1 HORMAD1 ADAMTSL4-AS2 GOLPH3L MIR6878 LOC124904417 MIR4257 MCL1 ENSA TARS2 ADAMTSL4-AS1 CTSS RPRD2 LOC107985203 ADAMTSL4 FALEC
          nsv6322618 Chr.1:150579001 - 150580100 on Build GRCh38 Deletion LOC107985203 MCL1
          nsv4057833 Chr.1:150439004 - 150716344 on Build GRCh38 Duplication ECM1 HORMAD1 ADAMTSL4-AS2 GOLPH3L MIR6878 LOC124904417 MIR4257 MCL1 ENSA TARS2 ADAMTSL4-AS1 RPRD2 LOC107985203 ADAMTSL4 FALEC

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          cell fate determination
          release of cytochrome c from mitochondria
          apoptotic process
          cellular response to DNA damage stimulus
          mitochondrial fusion
          intrinsic apoptotic signaling pathway in response to DNA damage
          negative regulation of autophagy
          cellular homeostasis
          cell differentiation
          response to cytokine
          regulation of apoptotic process
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of neuron apoptotic process
          transmembrane transport
          protein transmembrane transport
          extrinsic apoptotic signaling pathway in absence of ligand
          regulation of intracellular signal transduction
          positive regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
          negative regulation of anoikis
          regulation of apoptotic signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          protein binding
          protein transmembrane transporter activity
          channel activity
          protein heterodimerization activity
          BH domain binding
          BH3 domain binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline