Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00296806_cn
          See other LZTR1 CNV Assays ›
          Gene Symbol
          LZTR1
          Assay Reference Genome
          Location

          Chr.22:20983078 on build GRCh38
          Cytoband
          22q11.21
          Assay ID Hs00296806_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          8216

          Gene Name:

          leucine zipper like transcription regulator 1

          Gene Aliases:

          BTBD29, LZTR-1, NS10, SWNTS2

          Location:

          Chr.22:20982269-20999037 on Build GRCh38

          Assay Gene Location:

          Overlaps Exon 2 - Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          LZTR1 NM_006767.3 NP_006758.2
          AK296347.1
          AK310747.1
          BC026214.1 AAH26214.2
          CT841521.1 CAJ86451.1
          D38496.1 BAA07508.1
          DA194822.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3893441 Chr.22:20733858 - 21135629 on Build GRCh38 Gain+Loss P2RX6P MIR649 LZTR1 SERPIND1 LRRC74B THAP7 SLC7A4 BCRP2 P2RX6 CRKL THAP7-AS1 LOC101928891 TUBA3FP PI4KA SNAP29 AIFM3
          nsv1065617 Chr.22:20705383 - 21111546 on Build GRCh38 Gain P2RX6P MIR649 LZTR1 SERPIND1 LRRC74B THAP7 SLC7A4 BCRP2 P2RX6 CRKL THAP7-AS1 LOC101928891 TUBA3FP PI4KA SNAP29 AIFM3
          dgv7994n54 Chr.22:20559470 - 21111491 on Build GRCh38 Gain P2RX6P MIR649 LZTR1 SERPIND1 SMPD4P1 POM121L4P LRRC74B THAP7 SLC7A4 BCRP2 MED15 P2RX6 CRKL THAP7-AS1 LOC101928891 TUBA3FP TMEM191A PI4KA SNAP29 AIFM3
          nsv834142 Chr.22:20979844 - 21170235 on Build GRCh38 Gain P2RX6P MIR649 LZTR1 TUBA3FP FAM230B LRRC74B THAP7 SLC7A4 BCRP2 P2RX6 THAP7-AS1 AIFM3
          nsv471185 Chr.22:20977267 - 21033678 on Build GRCh38 Loss LZTR1 TUBA3FP THAP7 SLC7A4 P2RX6 THAP7-AS1 AIFM3
          nsv834141 Chr.22:20859455 - 21051952 on Build GRCh38 Loss P2RX6P MIR649 LZTR1 LRRC74B THAP7 SLC7A4 P2RX6 CRKL THAP7-AS1 LOC101928891 TUBA3FP SNAP29 AIFM3
          nsv828961 Chr.22:20980653 - 21023359 on Build GRCh38 Gain LZTR1 TUBA3FP THAP7 P2RX6 THAP7-AS1 AIFM3
          esv2763693 Chr.22:20581182 - 21111546 on Build GRCh38 Gain P2RX6P MIR649 LZTR1 SERPIND1 SMPD4P1 POM121L4P LRRC74B THAP7 SLC7A4 BCRP2 MED15 P2RX6 CRKL THAP7-AS1 LOC101928891 TUBA3FP TMEM191A PI4KA SNAP29 AIFM3
          esv3647304 Chr.22:20902577 - 21081487 on Build GRCh38 Loss P2RX6P LOC101928891 MIR649 LZTR1 TUBA3FP LRRC74B THAP7 SLC7A4 P2RX6 CRKL THAP7-AS1 AIFM3
          nsv588306 Chr.22:20963144 - 21111491 on Build GRCh38 Gain P2RX6P LOC101928891 MIR649 LZTR1 TUBA3FP LRRC74B THAP7 SLC7A4 BCRP2 P2RX6 THAP7-AS1 AIFM3
          nsv588305 Chr.22:20721841 - 21108064 on Build GRCh38 Loss P2RX6P MIR649 LZTR1 SERPIND1 LRRC74B THAP7 SLC7A4 BCRP2 P2RX6 CRKL THAP7-AS1 LOC101928891 TUBA3FP PI4KA SNAP29 AIFM3
          esv2678175 Chr.22:20691629 - 21284480 on Build GRCh38 Deletion P2RX6P MIR649 LZTR1 SERPIND1 POM121L4P GGT2 LRRC74B THAP7 SLC7A4 BCRP2 P2RX6 CRKL THAP7-AS1 LOC101928891 TUBA3FP FAM230B TMEM191A POM121L8P PI4KA SNAP29 AIFM3
          dgv7993n54 Chr.22:20346736 - 21111491 on Build GRCh38 Gain P2RX6P SCARF2 LZTR1 SERPIND1 SMPD4P1 P2RX6 THAP7-AS1 KLHL22 USP41 SNAP29 AIFM3 MIR649 POM121L4P LRRC74B THAP7 SLC7A4 BCRP2 MED15 CRKL ZNF74 LOC101928891 LOC101927859 TUBA3FP TMEM191A PI4KA
          dgv4483n100 Chr.22:20362634 - 21212930 on Build GRCh38 Gain P2RX6P SCARF2 LZTR1 SERPIND1 SMPD4P1 P2RX6 THAP7-AS1 KLHL22 USP41 SNAP29 AIFM3 MIR649 POM121L4P GGT2 LRRC74B THAP7 SLC7A4 BCRP2 MED15 CRKL ZNF74 LOC101928891 TUBA3FP FAM230B TMEM191A PI4KA
          nsv523162 Chr.22:20389246 - 21108064 on Build GRCh38 Loss P2RX6P MIR649 SCARF2 LZTR1 SERPIND1 SMPD4P1 POM121L4P LRRC74B THAP7 SLC7A4 BCRP2 MED15 P2RX6 CRKL THAP7-AS1 ZNF74 KLHL22 LOC101928891 TUBA3FP USP41 TMEM191A PI4KA SNAP29 AIFM3
          nsv828960 Chr.22:20953775 - 21057148 on Build GRCh38 Loss P2RX6P LOC101928891 MIR649 LZTR1 TUBA3FP LRRC74B THAP7 SLC7A4 P2RX6 THAP7-AS1 AIFM3

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          anatomical structure morphogenesis
          transcription factor activity, sequence-specific DNA binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline