Search
Search

Entrez Gene ID: | 390598 |
Gene Name: | SKI family transcriptional corepressor 1 |
Gene Aliases: |
CORL1, FUSSEL15, LBXCOR1 |
Location: |
Chr.15:67819704-67834561 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SKOR1 | NM_001258024.1 | NP_001244953.1 | ||
| XM_017022173.1 | XP_016877662.1 | |||
| AY669507.1 | AAV74188.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv827375 | Chr.15:67819001 - 67830434 on Build GRCh38 | Gain |
|
| nsv1578 | Chr.15:67827799 - 67862385 on Build GRCh38 | Insertion |
|
| nsv1041463 | Chr.15:67815528 - 67849977 on Build GRCh38 | Gain |
|
| nsv569803 | Chr.15:67585107 - 68124237 on Build GRCh38 | Gain |
|
Set Membership: |
|