Search
Search

Entrez Gene ID: | 796 |
Gene Name: | calcitonin related polypeptide alpha |
Gene Aliases: |
CALC1, CGRP, CGRP-I, CGRP1, CT, KC, PCT |
Location: |
Chr.11:14966668-14972361 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CALCA | NM_001033952.2 | 4 | 602 | NP_001029124.1 |
| NM_001033953.2 | NP_001029125.1 | |||
| NM_001741.2 | 4 | 578 | NP_001732.1 | |
| NR_125898.1 | ||||
| XM_017018283.1 | 4 | 653 | XP_016873772.1 | |
| XM_017018284.1 | 4 | 677 | XP_016873773.1 | |
| BC069684.1 | 4 | 578 | AAH69684.1 | |
| BC069704.1 | 4 | 578 | AAH69704.1 | |
| BC069760.1 | 4 | 578 | AAH69760.1 | |
| BC069778.1 | 4 | 578 | AAH69778.1 | |
| BC093753.1 | AAH93753.1 | |||
| BC101599.1 | AAI01600.1 | |||
| BC143562.1 | ||||
| BF942024.1 | 1 | 124 | ||
| K03512.1 | AAA52011.1 | |||
| M26095.1 | 4 | 561 | AAA35501.1 | |
| M64486.1 | AAA58403.1 | |||
| X00356.1 | 4 | 597 | CAA25103.1 | |
| X02330.1 | 2 | 383 | CAA26189.1 | |
| X03662.1 | 4 | 561 | CAA27299.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832071 | Chr.11:14854876 - 15027624 on Build GRCh38 | Loss |
|
| esv3578250 | Chr.11:14350967 - 14976647 on Build GRCh38 | Gain |
|
Set Membership: |
|