Search
Search

Entrez Gene ID: | 5498 |
Gene Name: | protoporphyrinogen oxidase |
Gene Aliases: |
PPO, V290M, VP, VPCO |
Location: |
Chr.1:161165728-161178013 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6332269 | Chr.1:160388767 - 162811116 on Build GRCh38 | Duplication |
|
| nsv4518029 | Chr.1:160848909 - 161416210 on Build GRCh38 | Duplication |
|
Set Membership: |
|