Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01020151_cn
          See other TALDO1 CNV Assays ›
          Gene Symbol
          TALDO1
          Assay Reference Genome
          Location

          Chr.11:763383 on build GRCh38
          Cytoband
          Assay ID Hs01020151_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          6888

          Gene Name:

          transaldolase 1

          Gene Aliases:

          TAL, TAL-H, TALDOR, TALH

          Location:

          Chr.11:747464-765012 on Build GRCh38

          Assay Gene Location:

          Within Exon 5
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          TALDO1 NM_006755.2 5 519 NP_006746.1
          AK313427.1 5 519 BAG36219.1
          BC010103.2 5 556 AAH10103.1
          BC018847.2 5 499 AAH18847.2
          HM005596.1 5 551 AEE61193.1
          L19437.2 5 551 AAB53943.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6450007 Chr.11:642551 - 894924 on Build GRCh38 Duplication CEND1 GATD1 CD151 PIDD1 LOC124900301 EPS8L2 POLR2L PNPLA2 CRACR2B SLC25A22 SNORA52 TMEM80 TSPAN4 RPLP2 CHID1 GATD1-DT DEAF1 PANO1 TALDO1
          nsv5508700 Chr.11:732123 - 768731 on Build GRCh38 Duplication GATD1 TALDO1
          esv2759794 Chr.11:409846 - 1336162 on Build GRCh38 Loss MIR6744 GATD1 EPS8L2 CRACR2B TSPAN4 CHID1 GATD1-DT DEAF1 LOC124902605 CEND1 DRD4 LRRC56 LINC02688 TOLLIP-DT PIDD1 LOC124900301 LOC143666 PNPLA2 LINC02708 RASSF7 MIR210HG MUC5B-AS1 PTDSS2 LMNTD2-AS1 CDHR5 MIR210 AP2A2 LOC124902805 SLC25A22 TOLLIP SIGIRR RNH1 PANO1 MUC5AC FAM99B CD151 IRF7 MUC2 LMNTD2 POLR2L SNORA52 TMEM80 SCT PHRF1 RPLP2 MUC6 ANO9 MUC5B HRAS TALDO1
          dgv182e199 Chr.11:349830 - 943372 on Build GRCh38 Deletion FAM99A AP2A2 LOC124902805 GATD1 EPS8L2 CRACR2B SLC25A22 TSPAN4 PKP3 KRTAP5-5 SIGIRR CHID1 B4GALNT4 GATD1-DT RNH1 DEAF1 PANO1 CEND1 DRD4 FAM99B LRRC56 CD151 PIDD1 IRF7 LMNTD2 LOC124900301 POLR2L LOC143666 PNPLA2 SNORA52 TMEM80 SCT PHRF1 RPLP2 LINC02708 KRTAP5-4 RASSF7 MIR210HG PTDSS2 LMNTD2-AS1 ANO9 CDHR5 HRAS TALDO1 MIR210
          esv3625077 Chr.11:732330 - 768730 on Build GRCh38 Gain GATD1 TALDO1
          nsv1159791 Chr.11:461793 - 909492 on Build GRCh38 Deletion LOC124902805 GATD1 EPS8L2 CRACR2B SLC25A22 TSPAN4 CHID1 GATD1-DT RNH1 DEAF1 PANO1 CEND1 DRD4 LRRC56 CD151 PIDD1 IRF7 LMNTD2 LOC124900301 POLR2L LOC143666 PNPLA2 SNORA52 TMEM80 SCT PHRF1 RPLP2 RASSF7 MIR210HG PTDSS2 LMNTD2-AS1 CDHR5 HRAS TALDO1 MIR210
          nsv6442745 Chr.11:699104 - 843498 on Build GRCh38 Duplication CEND1 GATD1 CD151 PIDD1 LOC124900301 EPS8L2 POLR2L PNPLA2 CRACR2B SLC25A22 SNORA52 TMEM80 TSPAN4 RPLP2 GATD1-DT DEAF1 PANO1 TALDO1
          nsv469923 Chr.11:378188 - 1302334 on Build GRCh38 Loss MIR6744 GATD1 EPS8L2 CRACR2B TSPAN4 CHID1 B4GALNT4 GATD1-DT DEAF1 LOC124902605 CEND1 DRD4 LRRC56 LINC02688 PIDD1 LOC124900301 LOC143666 PNPLA2 LINC02708 RASSF7 MIR210HG MUC5B-AS1 PTDSS2 LMNTD2-AS1 CDHR5 MIR210 FAM99A AP2A2 LOC124902805 SLC25A22 PKP3 TOLLIP SIGIRR RNH1 PANO1 MUC5AC FAM99B CD151 IRF7 MUC2 LMNTD2 POLR2L SNORA52 TMEM80 SCT PHRF1 RPLP2 MUC6 ANO9 MUC5B HRAS TALDO1
          nsv4210734 Chr.11:717201 - 926263 on Build GRCh38 Duplication CEND1 AP2A2 GATD1 CD151 PIDD1 LOC124900301 EPS8L2 POLR2L PNPLA2 CRACR2B SLC25A22 SNORA52 TSPAN4 RPLP2 CHID1 GATD1-DT PANO1 TALDO1
          esv3625081 Chr.11:756680 - 767369 on Build GRCh38 Gain GATD1 TALDO1
          nsv3221229 Chr.11:763165 - 764482 on Build GRCh38 Deletion TALDO1
          nsv552870 Chr.11:743813 - 810882 on Build GRCh38 Loss CEND1 RPLP2 GATD1 GATD1-DT PIDD1 LOC124900301 PANO1 SLC25A22 TALDO1
          nsv6443329 Chr.11:763172 - 764590 on Build GRCh38 Duplication TALDO1
          nsv951272 Chr.11:751201 - 911500 on Build GRCh38 Deletion CEND1 GATD1 CD151 PIDD1 LOC124900301 POLR2L PNPLA2 CRACR2B SLC25A22 SNORA52 TSPAN4 RPLP2 CHID1 GATD1-DT PANO1 TALDO1

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          fructose 6-phosphate metabolic process
          pentose-phosphate shunt
          pentose-phosphate shunt, non-oxidative branch
          glyceraldehyde-3-phosphate metabolic process
          sedoheptulose-7-phosphate:D-glyceraldehyde-3-phosphate glyceronetransferase activity
          protein binding
          transferase activity
          carbohydrate binding
          monosaccharide binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline