Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01072373_cn
          See other FLNC CNV Assays ›
          Gene Symbol
          FLNC
          Assay Reference Genome
          Location

          Chr.7:128855207 on build GRCh38
          Cytoband
          Assay ID Hs01072373_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          2318

          Gene Name:

          filamin C

          Gene Aliases:

          ABP-280, ABP280A, ABPA, ABPL, ARVC15, CMH26, FLN2, MFM5, MPD4, RCM5

          Location:

          Chr.7:128830406-128859272 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 42 - Exon 43
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          FLNC NM_001127487.2 NP_001120959.1
          NM_001458.5 NP_001449.3
          AB208865.1 BAD92102.1
          AB371585.1 BAG48314.1
          AF089841.1 AAD12245.1
          AF146692.1 AAF80245.1
          AJ012737.1 CAB46442.1

          Target Gene Details

          Entrez Gene ID:

          110806300

          Gene Name:

          FLNC antisense RNA 1

          Gene Aliases:

          -

          Location:

          Chr.7:128850162-128862626 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          FLNC-AS1 NR_149055.1
          DA760426.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv608411 Chr.7:128853335 - 128859025 on Build GRCh38 Loss FLNC-AS1 FLNC
          esv2670312 Chr.7:128499519 - 128999613 on Build GRCh38 Deletion GARIN1A FLNC ATP6V1F OPN1SW METTL2B CALU FLNC-AS1 GARIN1B KCP LINC01000 SPMIP1 CCDC136 TNPO3 IRF5
          nsv608409 Chr.7:128843173 - 128911686 on Build GRCh38 Loss FLNC-AS1 KCP FLNC SPMIP1 ATP6V1F
          nsv5914456 Chr.7:128499704 - 128999795 on Build GRCh38 Duplication GARIN1A FLNC ATP6V1F OPN1SW METTL2B CALU FLNC-AS1 GARIN1B KCP LINC01000 SPMIP1 CCDC136 TNPO3 IRF5
          esv22837 Chr.7:128774435 - 128953151 on Build GRCh38 Gain FLNC-AS1 KCP FLNC SPMIP1 CCDC136 ATP6V1F IRF5 OPN1SW
          nsv608408 Chr.7:128840161 - 128857116 on Build GRCh38 Gain FLNC-AS1 FLNC
          nsv5911338 Chr.7:128499612 - 128999703 on Build GRCh38 Deletion GARIN1A FLNC ATP6V1F OPN1SW METTL2B CALU FLNC-AS1 GARIN1B KCP LINC01000 SPMIP1 CCDC136 TNPO3 IRF5
          nsv517371 Chr.7:128832927 - 128933913 on Build GRCh38 Gain+Loss FLNC-AS1 KCP FLNC SPMIP1 ATP6V1F
          esv2735137 Chr.7:128499440 - 128999664 on Build GRCh38 Deletion GARIN1A FLNC ATP6V1F OPN1SW METTL2B CALU FLNC-AS1 GARIN1B KCP LINC01000 SPMIP1 CCDC136 TNPO3 IRF5
          dgv11611n54 Chr.7:128794846 - 128881377 on Build GRCh38 Loss FLNC-AS1 KCP FLNC SPMIP1 CCDC136 ATP6V1F
          dgv11612n54 Chr.7:128818595 - 128928906 on Build GRCh38 Loss FLNC-AS1 KCP FLNC SPMIP1 CCDC136 ATP6V1F
          nsv831125 Chr.7:128772651 - 128955697 on Build GRCh38 Loss CALU FLNC-AS1 KCP FLNC SPMIP1 CCDC136 TNPO3 ATP6V1F IRF5 OPN1SW
          nsv464713 Chr.7:128805212 - 128881377 on Build GRCh38 Loss FLNC-AS1 KCP FLNC SPMIP1 CCDC136 ATP6V1F
          dgv11614n54 Chr.7:128853623 - 128857116 on Build GRCh38 Loss FLNC-AS1 FLNC
          nsv949943 Chr.7:128827947 - 128862346 on Build GRCh38 Deletion FLNC-AS1 FLNC
          dgv11613n54 Chr.7:128837493 - 128857116 on Build GRCh38 Loss FLNC-AS1 FLNC
          nsv608410 Chr.7:128846465 - 128856742 on Build GRCh38 Loss FLNC-AS1 FLNC

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          actin cytoskeleton organization
          sarcomere organization
          muscle cell development
          actin binding
          protein binding
          cytoskeletal protein binding
          ankyrin binding
          identical protein binding
          actin filament binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline