Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01094109_cn
          See other NPY4R CNV Assays ›
          Gene Symbol
          NPY4R
          Assay Reference Genome
          Location

          Chr.10:47087300 on build 37
          Cytoband
          10q11.22b
          Assay ID Hs01094109_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5540

          Gene Name:

          neuropeptide Y receptor Y4

          Gene Aliases:

          NPY4-R, PP1, PPYR1, Y4

          Location:

          Chr.10:47083534-47088320 on Build 37

          Assay Gene Location:

          Within Exon 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          NPY4R NM_001278794.1 2 780 NP_001265723.1
          NM_005972.5 3 936 NP_005963.4
          AY649985.1 3 936 AAV68196.1
          AY649986.1 2 780 AAV68197.1
          BC096238.3 1 577 AAH96238.1
          BC099637.3 1 577 AAH99637.1
          CR541950.1 1 517 CAG46748.1
          U42387.1 2 602 AAB07759.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh 37
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv734n100 Chr.10:46527665 - 47121369 on Build 37 Gain SYT15 BMS1P1 PTPN20A NPY4R FRMPD2P2 GPRIN2 LINC00842 FAM35BP
          dgv819n100 Chr.10:47076675 - 47153919 on Build 37 Gain+Loss NPY4R LINC00842 HNRNPA1P33
          esv34875 Chr.10:46216415 - 47742094 on Build 37 Gain ANXA8L1 FAM25B AGAP4 NPY4R FAM35DP GPRIN2 FAM35BP FAM21C SYT15 BMS1P1 PTPN20A FRMPD2P2 BMS1P2 ANTXRL LINC00842 HNRNPA1P33
          dgv732n100 Chr.10:46247355 - 47743504 on Build 37 Gain ANXA8L1 FAM25B AGAP4 NPY4R FAM35DP GPRIN2 FAM35BP FAM21C SYT15 BMS1P1 PTPN20A FRMPD2P2 BMS1P2 ANTXRL LINC00842 HNRNPA1P33
          nsv1046878 Chr.10:47055642 - 47103613 on Build 37 Gain+Loss NPY4R LINC00842
          esv22204 Chr.10:46478317 - 47429141 on Build 37 Gain+Loss ANXA8L1 FAM25B NPY4R FAM35DP GPRIN2 FAM35BP SYT15 BMS1P1 PTPN20A FRMPD2P2 BMS1P2 LINC00842 HNRNPA1P33
          nsv470934 Chr.10:46990727 - 47703613 on Build 37 Gain ANXA8L1 FAM25B NPY4R FAM35DP BMS1P2 GPRIN2 ANTXRL LINC00842 HNRNPA1P33
          dgv765n100 Chr.10:46943377 - 47249575 on Build 37 Loss ANXA8L1 FAM25B SYT15 NPY4R BMS1P2 GPRIN2 LINC00842 HNRNPA1P33
          esv3333912 Chr.10:47072546 - 47133044 on Build 37 Duplication NPY4R LINC00842
          nsv1152446 Chr.10:46939299 - 47151600 on Build 37 Duplication SYT15 NPY4R GPRIN2 LINC00842 HNRNPA1P33
          nsv511437 Chr.10:47058530 - 47109348 on Build 37 Loss NPY4R LINC00842
          dgv740n100 Chr.10:46590219 - 47142310 on Build 37 Gain+Loss SYT15 BMS1P1 PTPN20A NPY4R FRMPD2P2 GPRIN2 LINC00842 FAM35BP HNRNPA1P33
          dgv223e212 Chr.10:46996612 - 47148814 on Build 37 Loss NPY4R GPRIN2 LINC00842 HNRNPA1P33
          dgv786n100 Chr.10:47000587 - 47113250 on Build 37 Gain NPY4R LINC00842
          dgv440n106 Chr.10:46914200 - 47153900 on Build 37 Duplication SYT15 NPY4R GPRIN2 LINC00842 FAM35BP HNRNPA1P33
          esv3891799 Chr.10:46961667 - 47149117 on Build 37 Gain SYT15 NPY4R GPRIN2 LINC00842 HNRNPA1P33
          nsv516485 Chr.10:46109533 - 48312291 on Build 37 Gain+Loss ANXA8L1 FAM25B AGAP4 NPY4R FAM35DP GPRIN2 BMS1P6 ZFAND4 FAM35BP FAM21C FAM25G SYT15 BMS1P1 PTPN20A FRMPD2P2 BMS1P2 CTSLP2 AGAP9 ANTXRL ANXA8L2 ANXA8 LINC00842 FAM21B HNRNPA1P33
          nsv1041060 Chr.10:47062299 - 47249575 on Build 37 Gain+Loss ANXA8L1 FAM25B NPY4R BMS1P2 LINC00842 HNRNPA1P33
          nsv550726 Chr.10:47041592 - 47153137 on Build 37 Loss NPY4R LINC00842 HNRNPA1P33
          esv3334625 Chr.10:47072646 - 47091244 on Build 37 Duplication NPY4R
          nsv1052733 Chr.10:47035615 - 47150458 on Build 37 Loss NPY4R LINC00842 HNRNPA1P33
          nsv821667 Chr.10:46977295 - 47632739 on Build 37 Gain ANXA8L1 FAM25B NPY4R FAM35DP BMS1P2 GPRIN2 LINC00842 HNRNPA1P33
          dgv767n100 Chr.10:46954483 - 47249575 on Build 37 Gain ANXA8L1 FAM25B SYT15 NPY4R BMS1P2 GPRIN2 LINC00842 HNRNPA1P33
          esv2736163 Chr.10:46802558 - 48992343 on Build 37 Deletion ANXA8L1 NPY4R FAM35DP GPRIN2 BMS1P6 FAM25G GDF10 RBP3 CTSLP2 AGAP9 ZNF488 FAM25B FRMPD2P1 FAM35BP SYT15 PTPN20B BMS1P2 BMS1P5 ANTXRL ANXA8L2 ANXA8 LINC00842 FAM21B GDF2 HNRNPA1P33
          dgv783n100 Chr.10:46989539 - 47153919 on Build 37 Gain NPY4R GPRIN2 LINC00842 HNRNPA1P33
          dgv755n100 Chr.10:46918172 - 47392320 on Build 37 Gain+Loss ANXA8L1 FAM25B SYT15 NPY4R FAM35DP BMS1P2 GPRIN2 LINC00842 FAM35BP HNRNPA1P33
          esv2750898 Chr.10:46045761 - 47593951 on Build 37 Loss ANXA8L1 FAM25B MARCH8 AGAP4 NPY4R FAM35DP GPRIN2 ZFAND4 FAM35BP FAM21C SYT15 BMS1P1 PTPN20A FRMPD2P2 BMS1P2 LINC00842 HNRNPA1P33
          dgv821n100 Chr.10:47087031 - 47153919 on Build 37 Gain NPY4R LINC00842 HNRNPA1P33
          esv2750899 Chr.10:46210751 - 47136996 on Build 37 Gain SYT15 BMS1P1 PTPN20A AGAP4 NPY4R FRMPD2P2 GPRIN2 LINC00842 FAM35BP HNRNPA1P33 FAM21C
          esv2759747 Chr.10:46021713 - 48481919 on Build 37 Gain+Loss ANXA8L1 NPY4R FAM35DP GPRIN2 BMS1P6 ZFAND4 FAM25G GDF10 RBP3 CTSLP2 AGAP9 ZNF488 FAM25B MARCH8 AGAP4 FAM35BP FAM21C SYT15 BMS1P1 PTPN20A FRMPD2P2 BMS1P2 ANTXRL ANXA8L2 ANXA8 LINC00842 FAM21B GDF2 HNRNPA1P33
          nsv1036434 Chr.10:47067469 - 47100644 on Build 37 Loss NPY4R LINC00842
          nsv947929 Chr.10:46975081 - 47132029 on Build 37 Duplication NPY4R GPRIN2 LINC00842
          dgv214e212 Chr.10:46287822 - 47692945 on Build 37 Gain ANXA8L1 FAM25B AGAP4 NPY4R FAM35DP GPRIN2 FAM35BP FAM21C SYT15 BMS1P1 PTPN20A FRMPD2P2 BMS1P2 ANTXRL LINC00842 HNRNPA1P33
          esv2632472 Chr.10:46952410 - 47149265 on Build 37 Gain SYT15 NPY4R GPRIN2 LINC00842 HNRNPA1P33
          dgv817n100 Chr.10:47063106 - 47249575 on Build 37 Loss ANXA8L1 FAM25B NPY4R BMS1P2 LINC00842 HNRNPA1P33
          dgv212e212 Chr.10:46256560 - 47149411 on Build 37 Gain SYT15 BMS1P1 PTPN20A AGAP4 NPY4R FRMPD2P2 GPRIN2 LINC00842 FAM35BP HNRNPA1P33 FAM21C
          dgv820n100 Chr.10:47078696 - 47121369 on Build 37 Loss NPY4R LINC00842
          dgv224e212 Chr.10:47045153 - 47109535 on Build 37 Gain NPY4R LINC00842
          nsv1075820 Chr.10:47008499 - 47149800 on Build 37 Duplication NPY4R LINC00842 HNRNPA1P33
          dgv803n100 Chr.10:47047587 - 47153919 on Build 37 Gain NPY4R LINC00842 HNRNPA1P33
          dgv31n111 Chr.10:47030900 - 47136996 on Build 37 Duplication NPY4R LINC00842 HNRNPA1P33
          nsv550725 Chr.10:47028331 - 47741882 on Build 37 Gain ANXA8L1 FAM25B NPY4R FAM35DP BMS1P2 ANTXRL LINC00842 HNRNPA1P33
          dgv745n100 Chr.10:46674168 - 47967299 on Build 37 Gain ANXA8L1 FAM25B NPY4R FAM35DP GPRIN2 FAM35BP SYT15 BMS1P1 FRMPD2P2 BMS1P2 ANTXRL ANXA8L2 LINC00842 FAM21B HNRNPA1P33
          dgv731n100 Chr.10:46148750 - 47148630 on Build 37 Gain SYT15 BMS1P1 PTPN20A AGAP4 NPY4R FRMPD2P2 GPRIN2 LINC00842 ZFAND4 FAM35BP HNRNPA1P33 FAM21C
          dgv774n100 Chr.10:46961423 - 47392320 on Build 37 Loss ANXA8L1 FAM25B SYT15 NPY4R FAM35DP BMS1P2 GPRIN2 LINC00842 HNRNPA1P33
          esv2736152 Chr.10:46698486 - 47330067 on Build 37 Deletion ANXA8L1 FAM25B SYT15 BMS1P1 NPY4R BMS1P2 GPRIN2 LINC00842 FAM35BP HNRNPA1P33
          dgv148n67 Chr.10:47000481 - 47149453 on Build 37 Gain NPY4R GPRIN2 LINC00842 HNRNPA1P33
          dgv228e212 Chr.10:47076674 - 48006310 on Build 37 Gain ANXA8L1 FAM25B NPY4R FAM35DP BMS1P2 ANTXRL ANXA8L2 LINC00842 FAM21B HNRNPA1P33
          dgv787n100 Chr.10:47003692 - 47743504 on Build 37 Gain+Loss ANXA8L1 FAM25B NPY4R FAM35DP BMS1P2 ANTXRL LINC00842 HNRNPA1P33
          nsv818758 Chr.10:46990728 - 47703613 on Build 37 Gain ANXA8L1 FAM25B NPY4R FAM35DP BMS1P2 GPRIN2 ANTXRL LINC00842 HNRNPA1P33
          nsv1039516 Chr.10:47029572 - 47103613 on Build 37 Gain NPY4R LINC00842
          esv3584617 Chr.10:46674180 - 47097403 on Build 37 Loss SYT15 BMS1P1 NPY4R FRMPD2P2 GPRIN2 LINC00842 FAM35BP
          dgv781n100 Chr.10:46989080 - 47153919 on Build 37 Loss NPY4R GPRIN2 LINC00842 HNRNPA1P33
          dgv738n100 Chr.10:46543845 - 47153919 on Build 37 Loss SYT15 BMS1P1 PTPN20A NPY4R FRMPD2P2 GPRIN2 LINC00842 FAM35BP HNRNPA1P33
          nsv951576 Chr.10:46940001 - 47150300 on Build 37 Duplication SYT15 NPY4R GPRIN2 LINC00842 HNRNPA1P33
          dgv227e212 Chr.10:47062985 - 47149411 on Build 37 Loss NPY4R LINC00842 HNRNPA1P33
          dgv791n100 Chr.10:47020164 - 47108045 on Build 37 Loss NPY4R LINC00842
          dgv409e59 Chr.10:46952346 - 47150344 on Build 37 Duplication SYT15 NPY4R GPRIN2 LINC00842 HNRNPA1P33
          esv3577105 Chr.10:47080360 - 47135671 on Build 37 Gain NPY4R LINC00842 HNRNPA1P33
          nsv831853 Chr.10:47067800 - 47222252 on Build 37 Gain ANXA8L1 FAM25B NPY4R LINC00842 HNRNPA1P33
          dgv813n100 Chr.10:47058887 - 47142310 on Build 37 Loss NPY4R LINC00842 HNRNPA1P33
          dgv792n100 Chr.10:47025092 - 47392320 on Build 37 Gain+Loss ANXA8L1 FAM25B NPY4R FAM35DP BMS1P2 LINC00842 HNRNPA1P33
          dgv220e212 Chr.10:46966534 - 47692945 on Build 37 Gain ANXA8L1 FAM25B SYT15 NPY4R FAM35DP BMS1P2 GPRIN2 ANTXRL LINC00842 HNRNPA1P33
          nsv1161943 Chr.10:46996905 - 47392332 on Build 37 Duplication ANXA8L1 FAM25B NPY4R FAM35DP BMS1P2 GPRIN2 LINC00842 HNRNPA1P33
          dgv147n67 Chr.10:46949255 - 47149453 on Build 37 Gain SYT15 NPY4R GPRIN2 LINC00842 HNRNPA1P33
          nsv8636 Chr.10:46677594 - 49272522 on Build 37 Gain+Loss ANXA8L1 NPY4R FAM35DP GPRIN2 BMS1P6 FAM25G GDF10 RBP3 CTSLP2 AGAP9 CTGLF12P ZNF488 FAM25B FRMPD2P1 FAM25C FAM35BP SYT15 BMS1P1 FRMPD2P2 PTPN20B BMS1P2 BMS1P5 ANTXRL ANXA8L2 ANXA8 LINC00842 FAM21B GDF2 HNRNPA1P33
          dgv811n100 Chr.10:47058780 - 47103613 on Build 37 Gain NPY4R LINC00842
          esv3578846 Chr.10:47063105 - 48174779 on Build 37 Loss ANXA8L1 FAM25B NPY4R FAM35DP BMS1P2 CTSLP2 ANTXRL ANXA8L2 LINC00842 FAM21B HNRNPA1P33
          esv3578852 Chr.10:47085737 - 47149411 on Build 37 Loss NPY4R LINC00842 HNRNPA1P33
          dgv801n100 Chr.10:47046997 - 47142310 on Build 37 Gain NPY4R LINC00842 HNRNPA1P33
          esv3382741 Chr.10:47083846 - 47091144 on Build 37 Duplication NPY4R
          nsv1044044 Chr.10:46977283 - 47138582 on Build 37 Gain+Loss NPY4R GPRIN2 LINC00842 HNRNPA1P33
          dgv773n100 Chr.10:46961233 - 47743504 on Build 37 Loss ANXA8L1 FAM25B SYT15 NPY4R FAM35DP BMS1P2 GPRIN2 ANTXRL LINC00842 HNRNPA1P33
          esv2857124 Chr.10:47072000 - 47148000 on Build 37 Duplication NPY4R LINC00842 HNRNPA1P33
          nsv436102 Chr.10:46680255 - 49378479 on Build 37 Deletion ANXA8L1 NPY4R FAM35DP GPRIN2 BMS1P6 FAM25G GDF10 RBP3 CTSLP2 AGAP9 CTGLF12P ZNF488 FAM25B FRMPD2P1 FAM25C FAM35BP SYT15 BMS1P1 FRMPD2P2 PTPN20B BMS1P2 BMS1P5 ANTXRL ANXA8L2 ANXA8 LINC00842 FAM21B GDF2 HNRNPA1P33 FRMPD2
          dgv776n100 Chr.10:46975048 - 47108045 on Build 37 Loss NPY4R GPRIN2 LINC00842

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Biological Process -

          Signal transduction Cell surface receptor mediated signal transduction G-protein mediated signaling

          Molecular Function -

          Receptor G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          digestion
          feeding behavior
          blood circulation
          peptide YY receptor activity
          pancreatic polypeptide receptor activity
          G-protein coupled receptor activity
          peptide binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline