Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Entrez Gene ID: | 6622 |
Gene Name: | synuclein alpha |
Gene Aliases: |
NACP, PARK1, PARK4, PD1 |
Location: |
Chr.4:89724099-89838324 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SNCA | NM_000345.3 | 6 | 910 | NP_000336.1 |
NM_001146054.1 | 6 | 906 | NP_001139526.1 | |
NM_001146055.1 | 6 | 717 | NP_001139527.1 | |
NM_007308.2 | 5 | 822 | NP_009292.1 | |
XM_011532208.2 | 6 | 787 | XP_011530510.1 | |
XM_017008562.1 | 6 | 781 | XP_016864051.1 | |
XM_017008563.1 | 6 | 807 | XP_016864052.1 | |
BC013293.2 | 6 | 1070 | AAH13293.1 | |
BC108275.1 | 6 | 864 | AAI08276.1 | |
D31839.1 | 4 | 576 | BAA06625.1 | |
JN709859.1 | 5 | 864 | ||
JN709860.1 | 4 | 780 | ||
JN709861.1 | 5 | 826 | ||
JN709862.1 | 5 | 868 | ||
JN709863.1 | 4 | 784 | ||
JN709864.1 | 5 | 633 | ||
JN709865.1 | 5 | 675 | ||
JN709866.1 | 4 | 591 | ||
JN709867.1 | 5 | 675 | ||
JN709868.1 | 5 | 717 | ||
JN709869.1 | 4 | 633 | ||
L08850.1 | 6 | 699 | AAA16117.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1012406 | Chr.4:89246630 - 90245636 on Build GRCh38 | Loss | SNCA MMRN1 SNCA-AS1 CCSER1 GPRIN3 |
nsv594806 | Chr.4:88621130 - 90539537 on Build GRCh38 | Gain | SNCA MMRN1 HERC3 SNCA-AS1 FAM13A CCSER1 TIGD2 FAM13A-AS1 NAP1L5 GPRIN3 |
Set Membership: |
Intragenic Exonic DGV Variation |