Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Entrez Gene ID: | 57716 |
Gene Name: | periaxin |
Gene Aliases: |
CMT4F |
Location: |
Chr.19:40393762-40414718 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRX | NM_020956.2 | 6 | 1846 | NP_066007.1 |
NM_181882.2 | 7 | 1197 | NP_870998.2 | |
XM_011527171.2 | 7 | 1189 | XP_011525473.1 | |
XM_017027046.1 | 3 | 1348 | XP_016882535.1 | |
XM_017027047.1 | 4 | 891 | XP_016882536.1 | |
AB046840.1 | 3 | 1327 | BAB13446.1 | |
AF321191.1 | 7 | 1197 | AAK19279.1 | |
AF321192.1 | 6 | 1846 | AAK19280.1 | |
BC067266.1 | 7 | 1199 | AAH67266.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833831 | Chr.19:40290833 - 40454958 on Build GRCh38 | Loss | LOC105372489 HIPK4 C19orf47 PLD3 PRX BLVRB MIR6796 SERTAD3 SERTAD1 |
Set Membership: |
Intragenic Exonic DGV Variation |