Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01200372_cn
          See other LENG8 CNV Assays ›
          Gene Symbol
          LENG8
          Assay Reference Genome
          Location

          Chr.19:54461809 on build GRCh38
          Cytoband
          19q13.42
          Assay ID Hs01200372_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          114823

          Gene Name:

          leukocyte receptor cluster member 8

          Gene Aliases:

          pp13842

          Location:

          Chr.19:54448719-54462046 on Build GRCh38

          Assay Gene Location:

          Within Exon 16
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          LENG8 NM_052925.3 16 3488 NP_443157.1
          XM_005278248.4 15 5723 XP_005278305.1
          XM_005278249.4 15 6175 XP_005278306.1
          XM_005278250.4 14 5594 XP_005278307.1
          XM_005278252.4 14 6064 XP_005278309.1
          XM_006722996.3 14 5599 XP_006723059.1
          XM_006722997.3 13 5488 XP_006723060.1
          XM_011526413.2 15 5662 XP_011524715.1
          XM_011526414.2 14 5612 XP_011524716.1
          XM_011526415.2 16 3479 XP_011524717.1
          AF211973.1 1 683
          AF211974.1 2 683
          AF211975.1 1 1801
          AF318338.1 1 982 AAL55845.1
          AL834532.1 4 1190 CAD39188.2
          BC028048.1 16 3439 AAH28048.1
          BM969015.1

          Target Gene Details

          Entrez Gene ID:

          94059

          Gene Name:

          leukocyte receptor cluster member 9

          Gene Aliases:

          -

          Location:

          Chr.19:54461796-54463711 on Build GRCh38

          Assay Gene Location:

          Overlaps - Exon 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          LENG9 NM_001301782.1 NP_001288711.1
          NM_198988.1 NP_945339.2
          BC015921.1 AAH15921.1
          CA450132.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv3663n100 Chr.19:54289334 - 54592227 on Build GRCh38 Gain LAIR2 KIR3DX1 LILRA2 LILRA5 CDC42EP5 LENG9 LENG8-AS1 LILRA4 LENG8 TTYH1 LAIR1
          nsv953617 Chr.19:54401196 - 54463717 on Build GRCh38 Deletion LENG9 LENG8-AS1 LENG8 TTYH1
          esv2758771 Chr.19:54163847 - 54954956 on Build GRCh38 Gain+Loss LAIR2 LILRA6 KIR3DL2 KIR2DL1 RPS9 CDC42EP5 LENG9 LENG8-AS1 NCR1 KIR2DS4 LILRB5 KIR2DL4 KIR3DL3 LILRP2 FCAR NLRP7 MIR4752 LILRB3 LENG8 KIR3DL1 LILRA2 LILRB1 LOC101928804 LILRA4 MBOAT7 LILRA1 LAIR1 KIR3DX1 LILRA5 TMC4 LOC102724300 MIR8061 TTYH1 TSEN34 LILRB4 KIR2DL3 LILRB2
          nsv833878 Chr.19:54268967 - 54518890 on Build GRCh38 Loss LAIR2 MIR4752 LILRA5 CDC42EP5 LENG9 LENG8-AS1 LILRA4 LENG8 TTYH1 LILRB2 LAIR1

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          mitophagy in response to mitochondrial depolarization
          xenophagy
          protein binding
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline