Search
Search

Entrez Gene ID: | 51540 |
Gene Name: | selenocysteine lyase |
Gene Aliases: |
SCL, hSCL |
Location: |
Chr.2:238060924-238099413 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCLY | NM_016510.5 | NP_057594.4 | ||
| AB209458.2 | BAD92695.1 | |||
| AF175767.1 | AAF36816.1 | |||
| AK293283.1 | ||||
| AK298409.1 | ||||
| AK301009.1 | ||||
| BC000586.2 | AAH00586.2 | |||
| BC007891.2 | AAH07891.1 | |||
| DN995828.1 |
Entrez Gene ID: | 100533179 |
Gene Name: | UBE2F-SCLY readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.2:237966945-238099413 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2F-SCLY | NR_037904.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss |
|
Set Membership: |
|