Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01213311_cn
          See other DUSP22 CNV Assays ›
          Gene Symbol
          DUSP22
          Assay Reference Genome
          Location

          Chr.6:348817 on build GRCh38
          Cytoband
          6p25.3
          Assay ID Hs01213311_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          56940

          Gene Name:

          dual specificity phosphatase 22

          Gene Aliases:

          JKAP, JSP-1, JSP1, LMW-DSP2, LMWDSP2, MKP-x, MKPX, VHX

          Location:

          Chr.6:292057-351355 on Build GRCh38

          Assay Gene Location:

          Within Exon 9
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          DUSP22 NM_001286555.1 7 967 NP_001273484.1
          NM_020185.4 NP_064570.1
          NR_104473.1 6 917
          NR_104474.1 6 884
          NR_104475.1 4 800
          XM_011514757.2 4 429 XP_011513059.1
          XM_017011062.1 XP_016866551.1
          AB208997.1 4 320 BAD92234.1
          AF165519.1 AAF86649.1
          AF424702.1 AAL18850.1
          AK000383.1 3 293
          AK074410.1 3 293
          AK128760.1 4 1288
          AK296402.1 7 537
          AK310653.1 5 990
          AY249859.1 AAP76376.1
          BC016844.1 AAH16844.1
          BC022847.2 AAH22847.1
          BX362191.2 4 581
          BX648953.1 4 4966
          DA950312.1 6 500
          DB177174.1 6 421

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1161330 Chr.6:323576 - 373453 on Build GRCh38 Deletion DUSP22
          esv2752087 Chr.6:165391 - 367109 on Build GRCh38 Gain DUSP22 LOC285766
          dgv991n67 Chr.6:291793 - 382505 on Build GRCh38 Gain DUSP22
          dgv1701e212 Chr.6:254254 - 385771 on Build GRCh38 Loss DUSP22
          dgv1159e214 Chr.6:327586 - 368712 on Build GRCh38 Gain DUSP22
          dgv990n67 Chr.6:254941 - 396689 on Build GRCh38 Gain DUSP22 IRF4
          dgv5878n100 Chr.6:252145 - 385772 on Build GRCh38 Loss DUSP22
          dgv5891n100 Chr.6:290586 - 366333 on Build GRCh38 Gain+Loss DUSP22
          nsv830568 Chr.6:232604 - 411249 on Build GRCh38 Gain+Loss DUSP22 IRF4
          dgv1696e212 Chr.6:253363 - 386419 on Build GRCh38 Gain DUSP22
          esv2421645 Chr.6:298700 - 379015 on Build GRCh38 Duplication DUSP22
          dgv5884n100 Chr.6:257341 - 447036 on Build GRCh38 Gain DUSP22 IRF4
          dgv140e203 Chr.6:257353 - 382897 on Build GRCh38 Gain+Loss DUSP22
          dgv1705e212 Chr.6:294124 - 385771 on Build GRCh38 Gain DUSP22
          dgv3438e59 Chr.6:256452 - 383750 on Build GRCh38 Duplication DUSP22
          dgv736n27 Chr.6:253861 - 394293 on Build GRCh38 Loss DUSP22 IRF4
          esv3890780 Chr.6:274847 - 381918 on Build GRCh38 Gain+Loss DUSP22
          dgv56e213 Chr.6:254283 - 381149 on Build GRCh38 Loss DUSP22
          esv29987 Chr.6:269735 - 349386 on Build GRCh38 Loss DUSP22
          nsv950754 Chr.6:341601 - 378900 on Build GRCh38 Duplication DUSP22
          dgv5897n100 Chr.6:323084 - 365708 on Build GRCh38 Loss DUSP22
          dgv5874n100 Chr.6:237215 - 365708 on Build GRCh38 Gain DUSP22
          nsv1026006 Chr.6:347166 - 361143 on Build GRCh38 Gain DUSP22
          esv3264495 Chr.6:256000 - 382000 on Build GRCh38 Duplication DUSP22
          nsv1028946 Chr.6:334825 - 404771 on Build GRCh38 Gain DUSP22 IRF4
          dgv5899n100 Chr.6:330692 - 385772 on Build GRCh38 Loss DUSP22
          dgv5888n100 Chr.6:279695 - 385772 on Build GRCh38 Loss DUSP22
          dgv5903n100 Chr.6:346085 - 361143 on Build GRCh38 Loss DUSP22
          esv3570714 Chr.6:294713 - 381137 on Build GRCh38 Loss DUSP22
          esv28111 Chr.6:255650 - 384973 on Build GRCh38 Gain+Loss DUSP22
          nsv428134 Chr.6:60001 - 653718 on Build GRCh38 Gain DUSP22 IRF4 LINC00266-3 LOC285766 EXOC2
          dgv5902n100 Chr.6:342894 - 385772 on Build GRCh38 Gain DUSP22
          dgv5901n100 Chr.6:334167 - 385772 on Build GRCh38 Gain DUSP22
          dgv5887n100 Chr.6:277348 - 385772 on Build GRCh38 Gain DUSP22
          esv3584838 Chr.6:330704 - 382559 on Build GRCh38 Loss DUSP22
          dgv5896n100 Chr.6:314950 - 385772 on Build GRCh38 Gain+Loss DUSP22
          nsv1148804 Chr.6:256499 - 382500 on Build GRCh38 Duplication DUSP22
          nsv820552 Chr.6:254458 - 396285 on Build GRCh38 Duplication DUSP22 IRF4
          nsv1161329 Chr.6:303121 - 360535 on Build GRCh38 Duplication DUSP22
          nsv1124934 Chr.6:257400 - 382500 on Build GRCh38 Duplication DUSP22
          nsv821644 Chr.6:182761 - 411478 on Build GRCh38 Gain DUSP22 IRF4 LOC285766
          nsv437476 Chr.6:337804 - 365074 on Build GRCh38 Loss DUSP22
          dgv5880n100 Chr.6:253272 - 385772 on Build GRCh38 Gain+Loss DUSP22
          dgv5890n100 Chr.6:285695 - 381137 on Build GRCh38 Gain+Loss DUSP22
          dgv5869n100 Chr.6:149649 - 395798 on Build GRCh38 Gain DUSP22 IRF4 LOC285766
          nsv1016277 Chr.6:320860 - 366333 on Build GRCh38 Gain DUSP22
          esv3607835 Chr.6:180946 - 734275 on Build GRCh38 Loss DUSP22 IRF4 LOC285766 HUS1B EXOC2
          nsv10799 Chr.6:255920 - 384864 on Build GRCh38 Gain+Loss DUSP22
          dgv5893n100 Chr.6:302294 - 365708 on Build GRCh38 Gain DUSP22
          dgv5892n100 Chr.6:294274 - 389630 on Build GRCh38 Gain DUSP22
          dgv10265n54 Chr.6:312247 - 374333 on Build GRCh38 Loss DUSP22
          dgv5877n100 Chr.6:249798 - 350608 on Build GRCh38 Loss DUSP22
          dgv5872n100 Chr.6:231934 - 397937 on Build GRCh38 Gain DUSP22 IRF4
          nsv600764 Chr.6:295956 - 383546 on Build GRCh38 Gain DUSP22
          nsv823390 Chr.6:295899 - 452604 on Build GRCh38 Gain DUSP22 IRF4
          dgv5889n100 Chr.6:280387 - 365708 on Build GRCh38 Gain DUSP22
          esv32812 Chr.6:236171 - 378927 on Build GRCh38 Loss DUSP22
          esv3890779 Chr.6:322474 - 381918 on Build GRCh38 Loss DUSP22
          dgv10266n54 Chr.6:348482 - 381918 on Build GRCh38 Loss DUSP22
          dgv10264n54 Chr.6:296257 - 381918 on Build GRCh38 Loss DUSP22
          nsv1020113 Chr.6:331800 - 361143 on Build GRCh38 Gain+Loss DUSP22
          nsv514340 Chr.6:257332 - 380527 on Build GRCh38 Gain DUSP22

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          inactivation of MAPK activity
          negative regulation of T cell mediated immunity
          protein dephosphorylation
          apoptotic process
          transforming growth factor beta receptor signaling pathway
          multicellular organism development
          cell proliferation
          peptidyl-tyrosine dephosphorylation
          regulation of cell proliferation
          positive regulation of JNK cascade
          negative regulation of T cell receptor signaling pathway
          negative regulation of T cell activation
          protein tyrosine phosphatase activity
          protein tyrosine/serine/threonine phosphatase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline