Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01253958_cn
          See other ARSD CNV Assays ›
          Gene Symbol
          ARSD
          Assay Reference Genome
          Location

          Chr.X:2904177 on build GRCh38
          Cytoband
          Assay ID Hs01253958_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          414

          Gene Name:

          arylsulfatase D

          Gene Aliases:

          ASD

          Location:

          Chr.X:2903972-2929339 on Build GRCh38

          Assay Gene Location:

          Within Exon 12
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          ARSD NM_001669.4 10 4940 NP_001660.2
          XM_005274514.3 9 4805 XP_005274571.1
          XM_005274515.3 10 5292 XP_005274572.1
          XM_047442108.1 10 4850 XP_047298064.1
          AI289224.1 1 224
          AK055239.1 1 1920
          HM005546.1 10 4952 AEE61143.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6139100 Chr.X:2838546 - 3174066 on Build GRCh38 Duplication ARSD-AS1 ARSF ARSH ARSL ARSD GYG2
          esv34985 Chr.X:2623614 - 3258451 on Build GRCh38 Gain ARSD-AS1 CD99 XG ARSF CD99P1 ARSH ARSL ARSD GYG2
          nsv5431814 Chr.X:2531036 - 2924507 on Build GRCh38 Duplication LINC00102 ARSD-AS1 CD99 XG MIR6089 CD99P1 ARSD GYG2
          esv2758855 Chr.X:1703008 - 3276324 on Build GRCh38 Gain ARSD-AS1 CD99 XG CD99P1 ARSH LINC01546 ARSL ARSD ZBED1 GYG2 LINC00102 MIR6089 ARSF DHRSX
          nsv5422252 Chr.X:2655643 - 2992295 on Build GRCh38 Duplication ARSD-AS1 CD99 XG CD99P1 ARSL ARSD GYG2
          esv32931 Chr.X:2897832 - 2918346 on Build GRCh38 Loss ARSD-AS1 ARSD
          nsv526612 Chr.X:2782116 - 2929092 on Build GRCh38 Loss ARSD-AS1 XG ARSD GYG2

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Process(es)

          arylsulfatase activity
          hydrolase activity
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline